Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR20001475.hg.1 |
ENST00000246090.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20001475.hg.1 |
ENST00000377805.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20001475.hg.1 |
ENST00000545418.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20001475.hg.1 |
NM_001014977.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20001475.hg.1 |
NM_001159495.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20001475.hg.1 |
NM_178477.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20001475.hg.1 |
TC20000110.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR20001474.hg.1 |
ENST00000246090.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20001474.hg.1 |
NM_178477.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20001474.hg.1 |
TC20000110.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006802.hg.1 |
ENST00000246090.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006802.hg.1 |
ENST00000246090.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006802.hg.1 |
ENST00000246090.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006802.hg.1 |
ENST00000246090.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006802.hg.1 |
ENST00000246090.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006802.hg.1 |
ENST00000377805.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006802.hg.1 |
ENST00000377805.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006802.hg.1 |
ENST00000377805.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006802.hg.1 |
ENST00000377805.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006802.hg.1 |
ENST00000427254.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006802.hg.1 |
ENST00000427254.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006802.hg.1 |
ENST00000427254.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006802.hg.1 |
ENST00000467330.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006802.hg.1 |
ENST00000467330.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006802.hg.1 |
ENST00000545418.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006802.hg.1 |
ENST00000545418.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006802.hg.1 |
ENST00000545418.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006802.hg.1 |
NM_001014977.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006802.hg.1 |
NM_001014977.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006802.hg.1 |
NM_001014977.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006802.hg.1 |
NM_001014977.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006802.hg.1 |
NM_001159495.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006802.hg.1 |
NM_001159495.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006802.hg.1 |
NM_001159495.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006802.hg.1 |
NM_178477.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006802.hg.1 |
NM_178477.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006802.hg.1 |
NM_178477.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006802.hg.1 |
NM_178477.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006802.hg.1 |
NM_178477.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006802.hg.1 |
ENST00000246090.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006802.hg.1 |
ENST00000246090.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006802.hg.1 |
ENST00000246090.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006802.hg.1 |
ENST00000246090.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006802.hg.1 |
ENST00000246090.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006802.hg.1 |
ENST00000377805.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006802.hg.1 |
ENST00000377805.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006802.hg.1 |
ENST00000377805.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006802.hg.1 |
ENST00000377805.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006802.hg.1 |
ENST00000427254.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006802.hg.1 |
ENST00000427254.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006802.hg.1 |
ENST00000427254.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006802.hg.1 |
ENST00000467330.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006802.hg.1 |
ENST00000467330.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006802.hg.1 |
ENST00000545418.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006802.hg.1 |
ENST00000545418.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006802.hg.1 |
ENST00000545418.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006802.hg.1 |
NM_001014977.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006802.hg.1 |
NM_001014977.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006802.hg.1 |
NM_001014977.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006802.hg.1 |
NM_001014977.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006802.hg.1 |
NM_001159495.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006802.hg.1 |
NM_001159495.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006802.hg.1 |
NM_001159495.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006802.hg.1 |
NM_178477.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006802.hg.1 |
NM_178477.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006802.hg.1 |
NM_178477.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006802.hg.1 |
NM_178477.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006802.hg.1 |
NM_178477.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000146550.hg.1 |
ENST00000246090.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000146550.hg.1 |
NM_178477.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000146550.hg.1 |
TC2000006802.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000146550.hg.1 |
ENST00000246090.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000146550.hg.1 |
NM_178477.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000146552.hg.1 |
ENST00000246090.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000146552.hg.1 |
ENST00000377805.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000146552.hg.1 |
ENST00000427254.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000146552.hg.1 |
ENST00000545418.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000146552.hg.1 |
NM_001014977.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000146552.hg.1 |
NM_001159495.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000146552.hg.1 |
NM_178477.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000146552.hg.1 |
TC2000006802.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000146552.hg.1 |
ENST00000246090.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000146552.hg.1 |
ENST00000377805.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000146552.hg.1 |
ENST00000427254.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000146552.hg.1 |
ENST00000545418.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000146552.hg.1 |
NM_001014977.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000146552.hg.1 |
NM_001159495.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000146552.hg.1 |
NM_178477.4 |
Refseq |