Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR13005888.hg.1 |
ENST00000266943.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13005888.hg.1 |
ENST00000380814.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13005888.hg.1 |
ENST00000475385.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13005888.hg.1 |
NM_001135919.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13005888.hg.1 |
NM_181785.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13005888.hg.1 |
TC13000518.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008468.hg.1 |
ENST00000266943.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008468.hg.1 |
ENST00000266943.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008468.hg.1 |
ENST00000266943.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008468.hg.1 |
ENST00000266943.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008468.hg.1 |
ENST00000266943.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008468.hg.1 |
ENST00000266943.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008468.hg.1 |
ENST00000266943.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008468.hg.1 |
ENST00000266943.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008468.hg.1 |
ENST00000266943.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008468.hg.1 |
ENST00000380814.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008468.hg.1 |
ENST00000380814.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008468.hg.1 |
ENST00000380814.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008468.hg.1 |
ENST00000380814.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008468.hg.1 |
ENST00000380814.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008468.hg.1 |
ENST00000380814.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008468.hg.1 |
ENST00000380814.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008468.hg.1 |
ENST00000380814.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008468.hg.1 |
ENST00000475385.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008468.hg.1 |
ENST00000475385.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008468.hg.1 |
ENST00000475385.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008468.hg.1 |
ENST00000475385.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008468.hg.1 |
ENST00000475385.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008468.hg.1 |
ENST00000475385.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008468.hg.1 |
NM_001135919.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008468.hg.1 |
NM_001135919.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008468.hg.1 |
NM_001135919.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008468.hg.1 |
NM_001135919.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008468.hg.1 |
NM_001135919.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008468.hg.1 |
NM_001135919.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008468.hg.1 |
NM_001135919.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008468.hg.1 |
NM_001135919.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008468.hg.1 |
NM_001135919.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008468.hg.1 |
NM_001135919.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008468.hg.1 |
NM_181785.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008468.hg.1 |
NM_181785.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008468.hg.1 |
NM_181785.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008468.hg.1 |
NM_181785.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008468.hg.1 |
NM_181785.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008468.hg.1 |
NM_181785.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008468.hg.1 |
NM_181785.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008468.hg.1 |
NM_181785.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008468.hg.1 |
NM_181785.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008468.hg.1 |
NM_181785.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008468.hg.1 |
NM_181785.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008468.hg.1 |
ENST00000266943.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008468.hg.1 |
ENST00000266943.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008468.hg.1 |
ENST00000266943.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008468.hg.1 |
ENST00000266943.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008468.hg.1 |
ENST00000266943.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008468.hg.1 |
ENST00000266943.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008468.hg.1 |
ENST00000266943.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008468.hg.1 |
ENST00000266943.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008468.hg.1 |
ENST00000266943.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008468.hg.1 |
ENST00000380814.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008468.hg.1 |
ENST00000380814.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008468.hg.1 |
ENST00000380814.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008468.hg.1 |
ENST00000380814.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008468.hg.1 |
ENST00000380814.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008468.hg.1 |
ENST00000380814.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008468.hg.1 |
ENST00000380814.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008468.hg.1 |
ENST00000380814.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008468.hg.1 |
ENST00000475385.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008468.hg.1 |
ENST00000475385.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008468.hg.1 |
ENST00000475385.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008468.hg.1 |
ENST00000475385.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008468.hg.1 |
ENST00000475385.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008468.hg.1 |
ENST00000475385.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008468.hg.1 |
NM_001135919.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008468.hg.1 |
NM_001135919.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008468.hg.1 |
NM_001135919.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008468.hg.1 |
NM_001135919.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008468.hg.1 |
NM_001135919.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008468.hg.1 |
NM_001135919.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008468.hg.1 |
NM_001135919.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008468.hg.1 |
NM_001135919.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008468.hg.1 |
NM_001135919.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008468.hg.1 |
NM_001135919.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008468.hg.1 |
NM_181785.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008468.hg.1 |
NM_181785.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008468.hg.1 |
NM_181785.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008468.hg.1 |
NM_181785.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008468.hg.1 |
NM_181785.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008468.hg.1 |
NM_181785.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008468.hg.1 |
NM_181785.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008468.hg.1 |
NM_181785.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008468.hg.1 |
NM_181785.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008468.hg.1 |
NM_181785.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008468.hg.1 |
NM_181785.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300157105.hg.1 |
ENST00000266943.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300157105.hg.1 |
ENST00000380814.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300157105.hg.1 |
ENST00000475385.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300157105.hg.1 |
NM_001135919.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300157105.hg.1 |
NM_181785.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300157105.hg.1 |
ENST00000266943.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300157105.hg.1 |
ENST00000380814.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300157105.hg.1 |
ENST00000475385.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300157105.hg.1 |
NM_001135919.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300157105.hg.1 |
NM_181785.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300157105.hg.1 |
TC1300008468.hg.1 |
Clariom D TC |