Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010579.hg.1 |
ENST00000367755.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010579.hg.1 |
ENST00000367755.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010579.hg.1 |
ENST00000367755.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010579.hg.1 |
ENST00000367755.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010579.hg.1 |
ENST00000367755.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010579.hg.1 |
ENST00000367755.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010579.hg.1 |
ENST00000367755.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010579.hg.1 |
ENST00000367755.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010579.hg.1 |
ENST00000367755.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010579.hg.1 |
ENST00000367755.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010579.hg.1 |
ENST00000367755.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010579.hg.1 |
ENST00000367755.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010579.hg.1 |
ENST00000367755.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010579.hg.1 |
ENST00000472784.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010579.hg.1 |
ENST00000472784.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010579.hg.1 |
ENST00000472784.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010579.hg.1 |
ENST00000472784.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010579.hg.1 |
ENST00000472784.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010579.hg.1 |
ENST00000478457.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010579.hg.1 |
ENST00000478457.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010579.hg.1 |
ENST00000478457.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010579.hg.1 |
ENST00000478457.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010579.hg.1 |
ENST00000479749.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010579.hg.1 |
ENST00000479749.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010579.hg.1 |
ENST00000479749.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010579.hg.1 |
ENST00000479749.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010579.hg.1 |
ENST00000479749.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010579.hg.1 |
ENST00000479749.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010579.hg.1 |
ENST00000479749.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010579.hg.1 |
ENST00000530212.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010579.hg.1 |
ENST00000530212.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010579.hg.1 |
ENST00000530212.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010579.hg.1 |
ENST00000534514.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010579.hg.1 |
ENST00000534514.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010579.hg.1 |
ENST00000534514.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010579.hg.1 |
ENST00000534514.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010579.hg.1 |
NM_001002294.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010579.hg.1 |
NM_001002294.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010579.hg.1 |
NM_001002294.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010579.hg.1 |
NM_001002294.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010579.hg.1 |
NM_001002294.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010579.hg.1 |
NM_001002294.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010579.hg.1 |
NM_001002294.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010579.hg.1 |
NM_001002294.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010579.hg.1 |
NM_001002294.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010579.hg.1 |
NM_001002294.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010579.hg.1 |
NM_001002294.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010579.hg.1 |
NM_001002294.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010579.hg.1 |
NM_001002294.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010579.hg.1 |
NM_006894.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010579.hg.1 |
NM_006894.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010579.hg.1 |
NM_006894.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010579.hg.1 |
NM_006894.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010579.hg.1 |
NM_006894.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010579.hg.1 |
NM_006894.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010579.hg.1 |
NM_006894.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010579.hg.1 |
NM_006894.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010579.hg.1 |
NM_006894.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010579.hg.1 |
NM_006894.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010579.hg.1 |
NM_006894.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010579.hg.1 |
NM_006894.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010579.hg.1 |
NM_006894.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01023047.hg.1 |
TC01001486.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01023047.hg.1 |
ENST00000367755.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01023047.hg.1 |
ENST00000392085.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01023047.hg.1 |
ENST00000538429.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01023047.hg.1 |
ENST00000542847.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01023047.hg.1 |
NM_001002294.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01023047.hg.1 |
NM_006894.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01023045.hg.1 |
TC01001486.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01023045.hg.1 |
ENST00000367755.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01023045.hg.1 |
ENST00000392085.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01023045.hg.1 |
ENST00000538429.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01023045.hg.1 |
ENST00000542847.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01023045.hg.1 |
NM_001002294.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01023045.hg.1 |
NM_006894.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100177862.hg.1 |
ENST00000367755.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100177862.hg.1 |
ENST00000472784.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100177862.hg.1 |
ENST00000478457.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100177862.hg.1 |
ENST00000479749.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100177862.hg.1 |
NM_001002294.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100177862.hg.1 |
NM_006894.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100177862.hg.1 |
ENST00000367755.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100177862.hg.1 |
ENST00000472784.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100177862.hg.1 |
ENST00000478457.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100177862.hg.1 |
ENST00000479749.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100177862.hg.1 |
NM_001002294.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100177862.hg.1 |
NM_006894.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100177862.hg.1 |
TC0100010579.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100010579.hg.1 |
ENST00000367755.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010579.hg.1 |
ENST00000367755.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010579.hg.1 |
ENST00000367755.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010579.hg.1 |
ENST00000367755.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010579.hg.1 |
ENST00000367755.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010579.hg.1 |
ENST00000367755.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010579.hg.1 |
ENST00000367755.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010579.hg.1 |
ENST00000367755.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010579.hg.1 |
ENST00000367755.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010579.hg.1 |
ENST00000367755.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010579.hg.1 |
ENST00000367755.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010579.hg.1 |
ENST00000367755.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010579.hg.1 |
ENST00000367755.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010579.hg.1 |
ENST00000472784.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010579.hg.1 |
ENST00000472784.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010579.hg.1 |
ENST00000472784.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010579.hg.1 |
ENST00000472784.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010579.hg.1 |
ENST00000472784.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010579.hg.1 |
ENST00000478457.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010579.hg.1 |
ENST00000478457.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010579.hg.1 |
ENST00000478457.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010579.hg.1 |
ENST00000478457.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010579.hg.1 |
ENST00000479749.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010579.hg.1 |
ENST00000479749.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010579.hg.1 |
ENST00000479749.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010579.hg.1 |
ENST00000479749.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010579.hg.1 |
ENST00000479749.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010579.hg.1 |
ENST00000479749.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010579.hg.1 |
ENST00000479749.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010579.hg.1 |
ENST00000530212.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010579.hg.1 |
ENST00000530212.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010579.hg.1 |
ENST00000530212.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010579.hg.1 |
ENST00000534514.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010579.hg.1 |
ENST00000534514.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010579.hg.1 |
ENST00000534514.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010579.hg.1 |
ENST00000534514.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010579.hg.1 |
NM_001002294.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010579.hg.1 |
NM_001002294.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010579.hg.1 |
NM_001002294.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010579.hg.1 |
NM_001002294.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010579.hg.1 |
NM_001002294.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010579.hg.1 |
NM_001002294.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010579.hg.1 |
NM_001002294.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010579.hg.1 |
NM_001002294.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010579.hg.1 |
NM_001002294.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010579.hg.1 |
NM_001002294.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010579.hg.1 |
NM_001002294.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010579.hg.1 |
NM_001002294.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010579.hg.1 |
NM_001002294.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010579.hg.1 |
NM_006894.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010579.hg.1 |
NM_006894.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010579.hg.1 |
NM_006894.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010579.hg.1 |
NM_006894.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010579.hg.1 |
NM_006894.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010579.hg.1 |
NM_006894.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010579.hg.1 |
NM_006894.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010579.hg.1 |
NM_006894.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010579.hg.1 |
NM_006894.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010579.hg.1 |
NM_006894.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010579.hg.1 |
NM_006894.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010579.hg.1 |
NM_006894.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010579.hg.1 |
NM_006894.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100177864.hg.1 |
ENST00000367755.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100177864.hg.1 |
ENST00000479749.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100177864.hg.1 |
NM_001002294.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100177864.hg.1 |
NM_006894.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100177864.hg.1 |
ENST00000367755.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100177864.hg.1 |
ENST00000479749.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100177864.hg.1 |
NM_001002294.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100177864.hg.1 |
NM_006894.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100177864.hg.1 |
TC0100010579.hg.1 |
Clariom D TC |