Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AHI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134830.1 | NP_001128302.1 | 20 - 21 | 3224 | 83 | |
NM_001134831.1 | NP_001128303.1 | 22 - 23 | 3371 | 83 | ||
NM_001134832.1 | NP_001128304.1 | 21 - 22 | 3286 | 83 | ||
NM_017651.4 | NP_060121.3 | 21 - 22 | 3286 | 83 | ||
XM_011535910.2 | XP_011534212.1 | 23 - 24 | 3515 | 83 | ||
XM_011535911.2 | XP_011534213.1 | 21 - 22 | 3245 | 83 | ||
XM_017010977.1 | XP_016866466.1 | 22 - 23 | 3490 | 83 | ||
XM_017010978.1 | XP_016866467.1 | 21 - 22 | 3188 | 83 | ||
XM_017010979.1 | XP_016866468.1 | 21 - 22 | 3188 | 83 | ||
XM_017010980.1 | XP_016866469.1 | 21 - 22 | 3188 | 83 | ||
XM_017010981.1 | XP_016866470.1 | 21 - 22 | 3323 | 83 | ||
XM_017010982.1 | XP_016866471.1 | 21 - 22 | 3188 | 83 | ||
XM_017010983.1 | XP_016866472.1 | 21 - 22 | 3188 | 83 | ||
XM_017010984.1 | XP_016866473.1 | 21 - 22 | 3188 | 83 | ||
GenBank mRNA | AJ459824.1 | - | 21 - 22 | 3266 | 83 | |
AJ459825.1 | - | 21 - 22 | 3266 | 83 | ||
AJ606362.1 | - | 20 - 21 | 3204 | 83 | ||
AK000076.1 | - | 13 - 14 | 1687 | 83 | ||
AK024085.1 | - | 9 - 10 | 1179 | 83 | ||
AK092262.1 | - | 20 - 21 | 3197 | 83 | ||
AK225210.1 | - | 13 - 14 | 1687 | 83 | ||
AL136797.1 | - | 19 - 20 | 3005 | 83 | ||
BC015019.1 | - | 11 - 12 | 1485 | 83 | ||
BC094800.1 | - | 22 - 23 | 3294 | 4823262 | 83 | |
DQ090887.1 | - | 22 - 23 | 3325 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons |