Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENPP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_006208.2 | NP_006199.2 | 14 - 15 | 1463 | 59 | |
XM_011535896.1 | XP_011534198.1 | 5 - 6 | 392 | 59 | ||
GenBank mRNA | AK314870.1 | - | 14 - 15 | 1290 | 59 | |
BC059375.1 | - | 14 - 15 | 1463 | 30341060 | 59 | |
BX537580.1 | - | 6 - 7 | 487 | 59 | ||
D12485.1 | - | 14 - 15 | 1459 | 59 | ||
M57736.1 | - | 14 - 15 | 1450 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons |