Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11029446.hg.1 |
ENST00000257359.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11029446.hg.1 |
ENST00000414575.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11029446.hg.1 |
NM_007037.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11029446.hg.1 |
TC11002452.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11029447.hg.1 |
ENST00000257359.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11029447.hg.1 |
ENST00000414575.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11029447.hg.1 |
NM_007037.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11029447.hg.1 |
TC11002452.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012834.hg.1 |
ENST00000257359.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012834.hg.1 |
ENST00000257359.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012834.hg.1 |
ENST00000257359.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012834.hg.1 |
ENST00000257359.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012834.hg.1 |
ENST00000257359.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012834.hg.1 |
ENST00000257359.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012834.hg.1 |
ENST00000257359.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012834.hg.1 |
ENST00000257359.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012834.hg.1 |
ENST00000257359.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012834.hg.1 |
ENST00000257359.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012834.hg.1 |
ENST00000257359.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012834.hg.1 |
ENST00000257359.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012834.hg.1 |
ENST00000257359.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012834.hg.1 |
ENST00000257359.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012834.hg.1 |
ENST00000531752.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012834.hg.1 |
ENST00000531752.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012834.hg.1 |
ENST00000531752.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012834.hg.1 |
ENST00000531752.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012834.hg.1 |
ENST00000531752.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012834.hg.1 |
ENST00000531752.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012834.hg.1 |
ENST00000531752.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012834.hg.1 |
ENST00000531752.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012834.hg.1 |
NM_007037.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012834.hg.1 |
NM_007037.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012834.hg.1 |
NM_007037.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012834.hg.1 |
NM_007037.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012834.hg.1 |
NM_007037.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012834.hg.1 |
NM_007037.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012834.hg.1 |
NM_007037.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012834.hg.1 |
NM_007037.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012834.hg.1 |
NM_007037.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012834.hg.1 |
NM_007037.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012834.hg.1 |
NM_007037.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012834.hg.1 |
NM_007037.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012834.hg.1 |
NM_007037.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100198502.hg.1 |
ENST00000257359.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100198502.hg.1 |
ENST00000531752.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100198502.hg.1 |
NM_007037.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100198502.hg.1 |
ENST00000257359.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100198502.hg.1 |
ENST00000531752.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100198502.hg.1 |
NM_007037.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100198502.hg.1 |
TC1100012834.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100012834.hg.1 |
ENST00000257359.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012834.hg.1 |
ENST00000257359.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012834.hg.1 |
ENST00000257359.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012834.hg.1 |
ENST00000257359.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012834.hg.1 |
ENST00000257359.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012834.hg.1 |
ENST00000257359.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012834.hg.1 |
ENST00000257359.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012834.hg.1 |
ENST00000257359.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012834.hg.1 |
ENST00000257359.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012834.hg.1 |
ENST00000257359.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012834.hg.1 |
ENST00000257359.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012834.hg.1 |
ENST00000257359.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012834.hg.1 |
ENST00000257359.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012834.hg.1 |
ENST00000257359.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012834.hg.1 |
ENST00000531752.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012834.hg.1 |
ENST00000531752.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012834.hg.1 |
ENST00000531752.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012834.hg.1 |
ENST00000531752.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012834.hg.1 |
ENST00000531752.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012834.hg.1 |
ENST00000531752.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012834.hg.1 |
ENST00000531752.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012834.hg.1 |
ENST00000531752.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012834.hg.1 |
NM_007037.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012834.hg.1 |
NM_007037.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012834.hg.1 |
NM_007037.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012834.hg.1 |
NM_007037.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012834.hg.1 |
NM_007037.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012834.hg.1 |
NM_007037.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012834.hg.1 |
NM_007037.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012834.hg.1 |
NM_007037.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012834.hg.1 |
NM_007037.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012834.hg.1 |
NM_007037.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012834.hg.1 |
NM_007037.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012834.hg.1 |
NM_007037.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012834.hg.1 |
NM_007037.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100198503.hg.1 |
ENST00000257359.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100198503.hg.1 |
ENST00000531752.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100198503.hg.1 |
NM_007037.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100198503.hg.1 |
ENST00000257359.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100198503.hg.1 |
ENST00000531752.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100198503.hg.1 |
NM_007037.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100198503.hg.1 |
TC1100012834.hg.1 |
Clariom D TC |