Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007736.hg.1 |
TC02000507.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007736.hg.1 |
ENST00000409013.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007736.hg.1 |
ENST00000409890.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007736.hg.1 |
NM_001135021.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007736.hg.1 |
NM_001135022.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007736.hg.1 |
NM_001135023.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007736.hg.1 |
NM_032213.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007757.hg.1 |
TC02000507.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007757.hg.1 |
ENST00000315658.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007757.hg.1 |
ENST00000393852.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007757.hg.1 |
ENST00000409013.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007757.hg.1 |
ENST00000409344.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007757.hg.1 |
ENST00000409890.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007757.hg.1 |
ENST00000428955.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007757.hg.1 |
NM_001135021.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007757.hg.1 |
NM_001135022.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007757.hg.1 |
NM_001135023.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007757.hg.1 |
NM_032213.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007744.hg.1 |
TC02000507.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007744.hg.1 |
ENST00000393852.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007744.hg.1 |
ENST00000409013.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007744.hg.1 |
ENST00000409344.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007744.hg.1 |
ENST00000409890.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007744.hg.1 |
ENST00000428955.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007744.hg.1 |
ENST00000490508.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007744.hg.1 |
NM_001135021.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007744.hg.1 |
NM_001135022.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007744.hg.1 |
NM_001135023.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007744.hg.1 |
NM_032213.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007737.hg.1 |
TC02000507.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007737.hg.1 |
ENST00000409013.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007737.hg.1 |
ENST00000409890.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007737.hg.1 |
ENST00000490508.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007737.hg.1 |
NM_001135021.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007737.hg.1 |
NM_001135022.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007737.hg.1 |
NM_001135023.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007737.hg.1 |
NM_032213.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007741.hg.1 |
TC02000507.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007741.hg.1 |
ENST00000393852.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007741.hg.1 |
ENST00000409013.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007741.hg.1 |
ENST00000409344.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007741.hg.1 |
ENST00000409890.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007741.hg.1 |
ENST00000428955.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007741.hg.1 |
ENST00000490508.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007741.hg.1 |
NM_001135021.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007741.hg.1 |
NM_001135022.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007741.hg.1 |
NM_001135023.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007741.hg.1 |
NM_032213.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007742.hg.1 |
TC02000507.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007742.hg.1 |
ENST00000393852.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007742.hg.1 |
ENST00000409013.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007742.hg.1 |
ENST00000409344.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007742.hg.1 |
ENST00000409890.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007742.hg.1 |
ENST00000428955.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007742.hg.1 |
ENST00000490508.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007742.hg.1 |
NM_001135021.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007742.hg.1 |
NM_001135022.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007742.hg.1 |
NM_001135023.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007742.hg.1 |
NM_032213.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007739.hg.1 |
TC02000507.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007739.hg.1 |
ENST00000409013.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007739.hg.1 |
ENST00000409890.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007739.hg.1 |
ENST00000490508.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007739.hg.1 |
NM_001135021.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007739.hg.1 |
NM_001135022.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007739.hg.1 |
NM_001135023.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007739.hg.1 |
NM_032213.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007740.hg.1 |
TC02000507.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007740.hg.1 |
ENST00000409013.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007740.hg.1 |
ENST00000409344.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007740.hg.1 |
ENST00000409890.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007740.hg.1 |
ENST00000490508.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007740.hg.1 |
NM_001135021.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007740.hg.1 |
NM_001135022.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007740.hg.1 |
NM_001135023.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007740.hg.1 |
NM_032213.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007745.hg.1 |
TC02000507.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007745.hg.1 |
ENST00000315658.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007745.hg.1 |
ENST00000393852.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007745.hg.1 |
ENST00000409013.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007745.hg.1 |
ENST00000409344.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007745.hg.1 |
ENST00000409890.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007745.hg.1 |
ENST00000428955.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007745.hg.1 |
ENST00000490508.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007745.hg.1 |
NM_001135021.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007745.hg.1 |
NM_001135022.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007745.hg.1 |
NM_001135023.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007745.hg.1 |
NM_032213.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007738.hg.1 |
TC02000507.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007738.hg.1 |
ENST00000409013.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007738.hg.1 |
ENST00000409890.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007738.hg.1 |
ENST00000490508.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007738.hg.1 |
NM_001135021.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007738.hg.1 |
NM_001135022.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007738.hg.1 |
NM_001135023.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007738.hg.1 |
NM_032213.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007743.hg.1 |
TC02000507.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007743.hg.1 |
ENST00000393852.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007743.hg.1 |
ENST00000409013.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007743.hg.1 |
ENST00000409344.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007743.hg.1 |
ENST00000409890.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007743.hg.1 |
ENST00000428955.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007743.hg.1 |
ENST00000490508.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007743.hg.1 |
NM_001135021.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007743.hg.1 |
NM_001135022.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007743.hg.1 |
NM_001135023.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007743.hg.1 |
NM_032213.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200157086.hg.1 |
ENST00000315658.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200157086.hg.1 |
ENST00000393852.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200157086.hg.1 |
ENST00000409013.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200157086.hg.1 |
ENST00000409331.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200157086.hg.1 |
ENST00000409344.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200157086.hg.1 |
ENST00000409890.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200157086.hg.1 |
ENST00000418268.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200157086.hg.1 |
ENST00000423095.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200157086.hg.1 |
ENST00000429764.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200157086.hg.1 |
ENST00000446464.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200157086.hg.1 |
ENST00000462396.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200157086.hg.1 |
ENST00000462891.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200157086.hg.1 |
ENST00000466467.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200157086.hg.1 |
ENST00000476734.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200157086.hg.1 |
NM_001135021.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200157086.hg.1 |
NM_001135022.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200157086.hg.1 |
NM_001135023.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200157086.hg.1 |
NM_032213.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200157086.hg.1 |
ENST00000315658.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200157086.hg.1 |
ENST00000393852.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200157086.hg.1 |
ENST00000409013.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200157086.hg.1 |
ENST00000409331.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200157086.hg.1 |
ENST00000409344.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200157086.hg.1 |
ENST00000409890.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200157086.hg.1 |
ENST00000418268.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200157086.hg.1 |
ENST00000423095.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200157086.hg.1 |
ENST00000429764.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200157086.hg.1 |
ENST00000446464.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200157086.hg.1 |
ENST00000462396.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200157086.hg.1 |
ENST00000462891.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200157086.hg.1 |
ENST00000466467.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200157086.hg.1 |
ENST00000476734.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200157086.hg.1 |
NM_001135021.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200157086.hg.1 |
NM_001135022.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200157086.hg.1 |
NM_001135023.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200157086.hg.1 |
NM_032213.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200157086.hg.1 |
TC0200008248.hg.1 |
Clariom D TC |