Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR16012618.hg.1 |
ENST00000261660.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16012618.hg.1 |
ENST00000381774.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16012618.hg.1 |
ENST00000433677.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16012618.hg.1 |
ENST00000539677.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16012618.hg.1 |
NM_001099455.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16012618.hg.1 |
NM_018340.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16012618.hg.1 |
TC16000879.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009446.hg.1 |
ENST00000261660.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009446.hg.1 |
ENST00000261660.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009446.hg.1 |
ENST00000261660.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009446.hg.1 |
ENST00000261660.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009446.hg.1 |
ENST00000261660.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009446.hg.1 |
ENST00000381774.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009446.hg.1 |
ENST00000381774.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009446.hg.1 |
ENST00000381774.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009446.hg.1 |
ENST00000381774.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009446.hg.1 |
ENST00000381774.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009446.hg.1 |
ENST00000381774.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009446.hg.1 |
ENST00000381774.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009446.hg.1 |
ENST00000381774.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009446.hg.1 |
ENST00000381774.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009446.hg.1 |
ENST00000381774.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009446.hg.1 |
ENST00000433677.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009446.hg.1 |
ENST00000433677.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009446.hg.1 |
ENST00000433677.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009446.hg.1 |
ENST00000433677.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009446.hg.1 |
ENST00000433677.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009446.hg.1 |
ENST00000433677.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009446.hg.1 |
ENST00000539677.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009446.hg.1 |
ENST00000539677.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009446.hg.1 |
ENST00000539677.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009446.hg.1 |
ENST00000539677.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009446.hg.1 |
NM_001099455.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009446.hg.1 |
NM_001099455.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009446.hg.1 |
NM_001099455.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009446.hg.1 |
NM_001099455.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009446.hg.1 |
NM_001099455.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009446.hg.1 |
NM_001099455.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009446.hg.1 |
NM_001099455.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009446.hg.1 |
NM_001099455.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009446.hg.1 |
NM_018340.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009446.hg.1 |
NM_018340.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009446.hg.1 |
NM_018340.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009446.hg.1 |
NM_018340.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009446.hg.1 |
NM_018340.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009446.hg.1 |
NM_018340.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009446.hg.1 |
NM_018340.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009446.hg.1 |
NM_018340.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009446.hg.1 |
NM_018340.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16012619.hg.1 |
ENST00000261660.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16012619.hg.1 |
ENST00000381774.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16012619.hg.1 |
ENST00000433677.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16012619.hg.1 |
ENST00000539677.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16012619.hg.1 |
NM_001099455.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16012619.hg.1 |
NM_018340.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16012619.hg.1 |
TC16000879.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1600009446.hg.1 |
ENST00000261660.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009446.hg.1 |
ENST00000261660.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009446.hg.1 |
ENST00000261660.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009446.hg.1 |
ENST00000261660.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009446.hg.1 |
ENST00000261660.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009446.hg.1 |
ENST00000381774.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009446.hg.1 |
ENST00000381774.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009446.hg.1 |
ENST00000381774.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009446.hg.1 |
ENST00000381774.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009446.hg.1 |
ENST00000381774.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009446.hg.1 |
ENST00000381774.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009446.hg.1 |
ENST00000381774.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009446.hg.1 |
ENST00000381774.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009446.hg.1 |
ENST00000381774.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009446.hg.1 |
ENST00000381774.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009446.hg.1 |
ENST00000433677.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009446.hg.1 |
ENST00000433677.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009446.hg.1 |
ENST00000433677.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009446.hg.1 |
ENST00000433677.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009446.hg.1 |
ENST00000433677.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009446.hg.1 |
ENST00000433677.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009446.hg.1 |
ENST00000539677.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009446.hg.1 |
ENST00000539677.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009446.hg.1 |
ENST00000539677.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009446.hg.1 |
ENST00000539677.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009446.hg.1 |
NM_001099455.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009446.hg.1 |
NM_001099455.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009446.hg.1 |
NM_001099455.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009446.hg.1 |
NM_001099455.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009446.hg.1 |
NM_001099455.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009446.hg.1 |
NM_001099455.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009446.hg.1 |
NM_001099455.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009446.hg.1 |
NM_001099455.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009446.hg.1 |
NM_018340.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009446.hg.1 |
NM_018340.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009446.hg.1 |
NM_018340.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009446.hg.1 |
NM_018340.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009446.hg.1 |
NM_018340.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009446.hg.1 |
NM_018340.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009446.hg.1 |
NM_018340.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009446.hg.1 |
NM_018340.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009446.hg.1 |
NM_018340.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600172366.hg.1 |
ENST00000261660.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600172366.hg.1 |
ENST00000381774.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600172366.hg.1 |
ENST00000433677.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600172366.hg.1 |
ENST00000539677.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600172366.hg.1 |
NM_001099455.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600172366.hg.1 |
NM_018340.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600172366.hg.1 |
ENST00000261660.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600172366.hg.1 |
ENST00000381774.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600172366.hg.1 |
ENST00000433677.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600172366.hg.1 |
ENST00000539677.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600172366.hg.1 |
NM_001099455.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600172366.hg.1 |
NM_018340.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600172366.hg.1 |
TC1600009446.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600172365.hg.1 |
ENST00000261660.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600172365.hg.1 |
ENST00000381774.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600172365.hg.1 |
ENST00000433677.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600172365.hg.1 |
ENST00000539677.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600172365.hg.1 |
NM_001099455.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600172365.hg.1 |
NM_018340.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600172365.hg.1 |
ENST00000261660.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600172365.hg.1 |
ENST00000381774.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600172365.hg.1 |
ENST00000433677.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600172365.hg.1 |
ENST00000539677.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600172365.hg.1 |
NM_001099455.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600172365.hg.1 |
NM_018340.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600172365.hg.1 |
TC1600009446.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600172367.hg.1 |
ENST00000261660.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600172367.hg.1 |
ENST00000381774.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600172367.hg.1 |
ENST00000433677.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600172367.hg.1 |
ENST00000539677.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600172367.hg.1 |
NM_001099455.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600172367.hg.1 |
NM_018340.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600172367.hg.1 |
ENST00000261660.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600172367.hg.1 |
ENST00000381774.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600172367.hg.1 |
ENST00000433677.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600172367.hg.1 |
ENST00000539677.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600172367.hg.1 |
NM_001099455.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600172367.hg.1 |
NM_018340.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600172367.hg.1 |
TC1600009446.hg.1 |
Clariom D TC |