Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002031.hg.1 |
ENST00000376835.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002031.hg.1 |
ENST00000480798.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002031.hg.1 |
NM_003650.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002031.hg.1 |
TC20000164.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007005.hg.1 |
ENST00000480798.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007005.hg.1 |
ENST00000480798.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007005.hg.1 |
ENST00000480798.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007005.hg.1 |
ENST00000480798.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007005.hg.1 |
ENST00000480798.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007005.hg.1 |
ENST00000480798.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007005.hg.1 |
ENST00000480798.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007005.hg.1 |
NM_003650.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007005.hg.1 |
NM_003650.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007005.hg.1 |
NM_003650.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007005.hg.1 |
NM_003650.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007005.hg.1 |
NM_003650.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007005.hg.1 |
NM_003650.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007005.hg.1 |
NM_003650.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007005.hg.1 |
ENST00000480798.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007005.hg.1 |
ENST00000480798.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007005.hg.1 |
ENST00000480798.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007005.hg.1 |
ENST00000480798.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007005.hg.1 |
ENST00000480798.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007005.hg.1 |
ENST00000480798.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007005.hg.1 |
ENST00000480798.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007005.hg.1 |
NM_003650.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007005.hg.1 |
NM_003650.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007005.hg.1 |
NM_003650.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007005.hg.1 |
NM_003650.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007005.hg.1 |
NM_003650.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007005.hg.1 |
NM_003650.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007005.hg.1 |
NM_003650.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000147773.hg.1 |
ENST00000480798.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000147773.hg.1 |
NM_003650.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000147773.hg.1 |
TC2000007005.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000147773.hg.1 |
ENST00000480798.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000147773.hg.1 |
NM_003650.3 |
Refseq |