Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSME4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_014614.2 | NP_055429.2 | 32 - 33 | 3743 | 145 | |
XM_006711969.1 | XP_006712032.1 | 31 - 32 | 3320 | 145 | ||
XM_011532705.2 | XP_011531007.1 | 33 - 34 | 3767 | 145 | ||
XM_011532706.2 | XP_011531008.1 | 33 - 34 | 3767 | 145 | ||
XM_011532707.2 | XP_011531009.1 | 32 - 33 | 3701 | 145 | ||
GenBank mRNA | AK025517.1 | - | 1 - 2 | 87 | 145 | |
AK026085.1 | - | 6 - 7 | 740 | 145 | ||
AY894754.1 | - | 31 - 32 | 3592 | 145 | ||
AY894755.1 | - | 13 - 14 | 1431 | 145 | ||
BC112169.1 | - | 31 - 32 | 3379 | 8327637 | 145 | |
BC113668.1 | - | 31 - 32 | 3379 | 8322720 | 145 | |
BC143739.1 | - | 31 - 32 | 3379 | 145 | ||
D38521.1 | - | 32 - 33 | 3553 | 145 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |