Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X011605.hg.1 |
TC0X000870.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X011605.hg.1 |
ENST00000380523.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X011605.hg.1 |
ENST00000398355.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X011605.hg.1 |
ENST00000460203.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X011605.hg.1 |
NM_001042479.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X011605.hg.1 |
NM_001042480.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X011605.hg.1 |
NM_017856.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00162413.hg.1 |
ENST00000332885.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00162413.hg.1 |
ENST00000380523.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00162413.hg.1 |
ENST00000398355.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00162413.hg.1 |
ENST00000460203.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00162413.hg.1 |
ENST00000477386.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00162413.hg.1 |
NM_001042479.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00162413.hg.1 |
NM_001042480.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00162413.hg.1 |
NM_017856.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00162413.hg.1 |
ENST00000332885.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00162413.hg.1 |
ENST00000380523.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00162413.hg.1 |
ENST00000398355.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00162413.hg.1 |
ENST00000460203.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00162413.hg.1 |
ENST00000477386.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00162413.hg.1 |
NM_001042479.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00162413.hg.1 |
NM_001042480.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00162413.hg.1 |
NM_017856.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00162413.hg.1 |
TC0X00009107.hg.1 |
Clariom D TC |