Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR16013653.hg.1 |
ENST00000268389.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16013653.hg.1 |
NM_005849.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16013653.hg.1 |
TC16000939.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009683.hg.1 |
ENST00000268389.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009683.hg.1 |
ENST00000268389.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009683.hg.1 |
ENST00000268389.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009683.hg.1 |
ENST00000268389.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009683.hg.1 |
ENST00000268389.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009683.hg.1 |
ENST00000268389.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009683.hg.1 |
ENST00000268389.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009683.hg.1 |
ENST00000268389.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009683.hg.1 |
ENST00000268389.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009683.hg.1 |
ENST00000268389.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009683.hg.1 |
ENST00000565499.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009683.hg.1 |
ENST00000565499.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009683.hg.1 |
ENST00000565499.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009683.hg.1 |
ENST00000565499.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009683.hg.1 |
ENST00000569602.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009683.hg.1 |
ENST00000569602.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009683.hg.1 |
ENST00000569602.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009683.hg.1 |
NM_005849.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009683.hg.1 |
NM_005849.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009683.hg.1 |
NM_005849.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009683.hg.1 |
NM_005849.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009683.hg.1 |
NM_005849.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009683.hg.1 |
NM_005849.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009683.hg.1 |
NM_005849.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009683.hg.1 |
NM_005849.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009683.hg.1 |
NM_005849.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600174479.hg.1 |
ENST00000268389.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600174479.hg.1 |
NM_005849.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600174479.hg.1 |
ENST00000268389.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600174479.hg.1 |
NM_005849.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600174479.hg.1 |
TC1600009683.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1600009683.hg.1 |
ENST00000268389.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009683.hg.1 |
ENST00000268389.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009683.hg.1 |
ENST00000268389.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009683.hg.1 |
ENST00000268389.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009683.hg.1 |
ENST00000268389.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009683.hg.1 |
ENST00000268389.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009683.hg.1 |
ENST00000268389.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009683.hg.1 |
ENST00000268389.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009683.hg.1 |
ENST00000268389.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009683.hg.1 |
ENST00000268389.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009683.hg.1 |
ENST00000565499.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009683.hg.1 |
ENST00000565499.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009683.hg.1 |
ENST00000565499.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009683.hg.1 |
ENST00000565499.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009683.hg.1 |
ENST00000569602.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009683.hg.1 |
ENST00000569602.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009683.hg.1 |
ENST00000569602.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009683.hg.1 |
NM_005849.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009683.hg.1 |
NM_005849.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009683.hg.1 |
NM_005849.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009683.hg.1 |
NM_005849.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009683.hg.1 |
NM_005849.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009683.hg.1 |
NM_005849.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009683.hg.1 |
NM_005849.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009683.hg.1 |
NM_005849.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009683.hg.1 |
NM_005849.3 |
Refseq |