Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012155.hg.1 |
ENST00000283871.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012155.hg.1 |
ENST00000283871.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012155.hg.1 |
ENST00000283871.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012155.hg.1 |
ENST00000283871.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012155.hg.1 |
ENST00000283871.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012155.hg.1 |
ENST00000283871.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012155.hg.1 |
ENST00000283871.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012155.hg.1 |
ENST00000283871.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012155.hg.1 |
ENST00000283871.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012155.hg.1 |
ENST00000283871.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012155.hg.1 |
ENST00000283871.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012155.hg.1 |
ENST00000283871.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012155.hg.1 |
ENST00000283871.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012155.hg.1 |
ENST00000283871.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012155.hg.1 |
ENST00000283871.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012155.hg.1 |
ENST00000283871.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012155.hg.1 |
ENST00000283871.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012155.hg.1 |
ENST00000283871.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012155.hg.1 |
ENST00000283871.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012155.hg.1 |
ENST00000283871.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012155.hg.1 |
ENST00000283871.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012155.hg.1 |
ENST00000283871.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012155.hg.1 |
ENST00000466528.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012155.hg.1 |
ENST00000466528.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012155.hg.1 |
ENST00000466528.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012155.hg.1 |
ENST00000466528.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012155.hg.1 |
ENST00000470321.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012155.hg.1 |
ENST00000470321.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012155.hg.1 |
ENST00000470321.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012155.hg.1 |
ENST00000470321.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012155.hg.1 |
ENST00000470321.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012155.hg.1 |
ENST00000470321.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012155.hg.1 |
ENST00000475447.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012155.hg.1 |
ENST00000475447.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012155.hg.1 |
ENST00000475447.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012155.hg.1 |
ENST00000475447.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012155.hg.1 |
ENST00000475447.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012155.hg.1 |
ENST00000476082.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012155.hg.1 |
ENST00000476082.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012155.hg.1 |
ENST00000476082.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012155.hg.1 |
ENST00000476082.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012155.hg.1 |
ENST00000476082.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012155.hg.1 |
ENST00000476082.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012155.hg.1 |
ENST00000476082.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012155.hg.1 |
ENST00000480862.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012155.hg.1 |
ENST00000480862.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012155.hg.1 |
ENST00000480862.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012155.hg.1 |
ENST00000480862.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012155.hg.1 |
ENST00000480862.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012155.hg.1 |
ENST00000480862.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012155.hg.1 |
ENST00000485313.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012155.hg.1 |
ENST00000485313.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012155.hg.1 |
ENST00000485313.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012155.hg.1 |
ENST00000485313.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012155.hg.1 |
ENST00000485313.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012155.hg.1 |
ENST00000485313.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012155.hg.1 |
ENST00000485313.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012155.hg.1 |
ENST00000488183.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012155.hg.1 |
ENST00000488183.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012155.hg.1 |
ENST00000488183.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012155.hg.1 |
ENST00000488183.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012155.hg.1 |
ENST00000488183.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012155.hg.1 |
ENST00000488183.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012155.hg.1 |
ENST00000488183.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012155.hg.1 |
ENST00000488183.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012155.hg.1 |
ENST00000492108.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012155.hg.1 |
ENST00000492108.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012155.hg.1 |
ENST00000492108.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012155.hg.1 |
ENST00000492108.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012155.hg.1 |
ENST00000492108.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012155.hg.1 |
ENST00000492108.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012155.hg.1 |
ENST00000492108.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012155.hg.1 |
ENST00000494453.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012155.hg.1 |
ENST00000494453.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012155.hg.1 |
ENST00000494453.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012155.hg.1 |
ENST00000494453.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012155.hg.1 |
ENST00000494453.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012155.hg.1 |
ENST00000494453.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012155.hg.1 |
NM_000187.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012155.hg.1 |
NM_000187.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012155.hg.1 |
NM_000187.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012155.hg.1 |
NM_000187.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012155.hg.1 |
NM_000187.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012155.hg.1 |
NM_000187.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012155.hg.1 |
NM_000187.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012155.hg.1 |
NM_000187.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012155.hg.1 |
NM_000187.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012155.hg.1 |
NM_000187.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012155.hg.1 |
NM_000187.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012155.hg.1 |
NM_000187.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012155.hg.1 |
NM_000187.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012155.hg.1 |
NM_000187.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012155.hg.1 |
NM_000187.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012155.hg.1 |
NM_000187.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012155.hg.1 |
NM_000187.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012155.hg.1 |
NM_000187.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012155.hg.1 |
NM_000187.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012155.hg.1 |
NM_000187.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012155.hg.1 |
NM_000187.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012155.hg.1 |
NM_000187.3 |
Refseq |