Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR14015219.hg.1 |
ENST00000298352.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14015219.hg.1 |
NM_021257.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14015219.hg.1 |
TC14001341.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009776.hg.1 |
ENST00000298352.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009776.hg.1 |
ENST00000298352.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009776.hg.1 |
ENST00000298352.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009776.hg.1 |
ENST00000298352.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009776.hg.1 |
ENST00000298352.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009776.hg.1 |
ENST00000298352.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009776.hg.1 |
ENST00000298352.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009776.hg.1 |
NM_021257.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009776.hg.1 |
NM_021257.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009776.hg.1 |
NM_021257.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009776.hg.1 |
NM_021257.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009776.hg.1 |
NM_021257.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009776.hg.1 |
NM_021257.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009776.hg.1 |
NM_021257.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14015217.hg.1 |
ENST00000298352.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14015217.hg.1 |
NM_021257.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14015217.hg.1 |
TC14001341.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR14015218.hg.1 |
ENST00000298352.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14015218.hg.1 |
NM_021257.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14015218.hg.1 |
TC14001341.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR14015216.hg.1 |
ENST00000298352.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14015216.hg.1 |
NM_021257.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14015216.hg.1 |
TC14001341.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1400009776.hg.1 |
ENST00000298352.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009776.hg.1 |
ENST00000298352.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009776.hg.1 |
ENST00000298352.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009776.hg.1 |
ENST00000298352.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009776.hg.1 |
ENST00000298352.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009776.hg.1 |
ENST00000298352.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009776.hg.1 |
ENST00000298352.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009776.hg.1 |
NM_021257.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009776.hg.1 |
NM_021257.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009776.hg.1 |
NM_021257.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009776.hg.1 |
NM_021257.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009776.hg.1 |
NM_021257.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009776.hg.1 |
NM_021257.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009776.hg.1 |
NM_021257.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400172075.hg.1 |
ENST00000298352.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400172075.hg.1 |
NM_021257.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400172075.hg.1 |
ENST00000298352.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400172075.hg.1 |
NM_021257.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400172075.hg.1 |
TC1400009776.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400172078.hg.1 |
ENST00000298352.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400172078.hg.1 |
NM_021257.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400172078.hg.1 |
ENST00000298352.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400172078.hg.1 |
NM_021257.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400172078.hg.1 |
TC1400009776.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400172077.hg.1 |
ENST00000298352.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400172077.hg.1 |
NM_021257.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400172077.hg.1 |
ENST00000298352.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400172077.hg.1 |
NM_021257.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400172077.hg.1 |
TC1400009776.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400172079.hg.1 |
ENST00000298352.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400172079.hg.1 |
NM_021257.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400172079.hg.1 |
ENST00000298352.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400172079.hg.1 |
NM_021257.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400172079.hg.1 |
TC1400009776.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400172076.hg.1 |
ENST00000298352.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400172076.hg.1 |
NM_021257.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400172076.hg.1 |
ENST00000298352.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400172076.hg.1 |
NM_021257.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400172076.hg.1 |
TC1400009776.hg.1 |
Clariom D TC |