Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19006481.hg.1 |
ENST00000262625.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19006481.hg.1 |
ENST00000341658.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19006481.hg.1 |
ENST00000347900.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19006481.hg.1 |
ENST00000360202.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19006481.hg.1 |
NM_032123.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19006481.hg.1 |
NM_199179.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19006481.hg.1 |
NM_199180.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19006481.hg.1 |
TC19000487.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19006480.hg.1 |
ENST00000262625.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19006480.hg.1 |
ENST00000270294.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19006480.hg.1 |
ENST00000341658.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19006480.hg.1 |
ENST00000347900.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19006480.hg.1 |
ENST00000360202.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19006480.hg.1 |
NM_032123.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19006480.hg.1 |
NM_199179.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19006480.hg.1 |
NM_199180.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19006480.hg.1 |
TC19000487.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1900158727.hg.1 |
ENST00000262625.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900158727.hg.1 |
ENST00000347900.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900158727.hg.1 |
ENST00000360202.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900158727.hg.1 |
ENST00000592409.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900158727.hg.1 |
NM_032123.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900158727.hg.1 |
NM_199179.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900158727.hg.1 |
NM_199180.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900158727.hg.1 |
TC1900007887.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900158727.hg.1 |
ENST00000262625.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900158727.hg.1 |
ENST00000347900.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900158727.hg.1 |
ENST00000360202.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900158727.hg.1 |
ENST00000592409.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900158727.hg.1 |
NM_032123.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900158727.hg.1 |
NM_199179.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900158727.hg.1 |
NM_199180.3 |
Refseq |