Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_003987.4 | NP_003978.3 | 6 - 7 | 1242 | 71 | |
NM_003990.4 | NP_003981.3 | 6 - 7 | 1242 | 71 | ||
XM_017016281.1 | XP_016871770.1 | 8 - 9 | 3653 | 71 | ||
XM_017016282.1 | XP_016871771.1 | 8 - 9 | 3653 | 71 | ||
XM_017016283.1 | XP_016871772.1 | 8 - 9 | 3653 | 71 | ||
XM_017016284.1 | XP_016871773.1 | 7 - 8 | 3554 | 71 | ||
XM_017016285.1 | XP_016871774.1 | 7 - 8 | 3554 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Homo sapiens Probe spans exons |