Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10012135.hg.1 |
ENST00000381344.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10012135.hg.1 |
NM_004508.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10012135.hg.1 |
TC10000967.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10012137.hg.1 |
ENST00000381344.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10012137.hg.1 |
NM_004508.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10012137.hg.1 |
TC10000967.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009536.hg.1 |
ENST00000381344.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009536.hg.1 |
ENST00000381344.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009536.hg.1 |
ENST00000381344.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009536.hg.1 |
ENST00000381344.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009536.hg.1 |
ENST00000381344.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009536.hg.1 |
ENST00000381344.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009536.hg.1 |
ENST00000381344.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009536.hg.1 |
ENST00000381344.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009536.hg.1 |
ENST00000381344.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009536.hg.1 |
ENST00000381344.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009536.hg.1 |
ENST00000381344.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009536.hg.1 |
ENST00000381344.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009536.hg.1 |
ENST00000427898.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009536.hg.1 |
ENST00000427898.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009536.hg.1 |
ENST00000427898.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009536.hg.1 |
ENST00000427898.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009536.hg.1 |
ENST00000427898.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009536.hg.1 |
ENST00000429642.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009536.hg.1 |
ENST00000429642.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009536.hg.1 |
ENST00000429642.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009536.hg.1 |
ENST00000429642.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009536.hg.1 |
ENST00000429642.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009536.hg.1 |
ENST00000429642.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009536.hg.1 |
ENST00000482091.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009536.hg.1 |
ENST00000482091.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009536.hg.1 |
ENST00000482091.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009536.hg.1 |
ENST00000482091.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009536.hg.1 |
ENST00000482091.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009536.hg.1 |
ENST00000491735.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009536.hg.1 |
ENST00000491735.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009536.hg.1 |
ENST00000491735.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009536.hg.1 |
ENST00000491735.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009536.hg.1 |
ENST00000491735.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009536.hg.1 |
ENST00000491735.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009536.hg.1 |
ENST00000491735.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009536.hg.1 |
ENST00000491735.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009536.hg.1 |
NM_004508.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009536.hg.1 |
NM_004508.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009536.hg.1 |
NM_004508.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009536.hg.1 |
NM_004508.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009536.hg.1 |
NM_004508.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009536.hg.1 |
NM_004508.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009536.hg.1 |
NM_004508.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009536.hg.1 |
NM_004508.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009536.hg.1 |
NM_004508.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009536.hg.1 |
NM_004508.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000165148.hg.1 |
ENST00000381344.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000165148.hg.1 |
NM_004508.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000165148.hg.1 |
ENST00000381344.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000165148.hg.1 |
NM_004508.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000165148.hg.1 |
TC1000009536.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000165149.hg.1 |
ENST00000381344.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000165149.hg.1 |
ENST00000491735.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000165149.hg.1 |
NM_004508.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000165149.hg.1 |
ENST00000381344.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000165149.hg.1 |
ENST00000491735.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000165149.hg.1 |
NM_004508.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000165149.hg.1 |
TC1000009536.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000165152.hg.1 |
ENST00000381344.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000165152.hg.1 |
ENST00000427898.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000165152.hg.1 |
ENST00000429642.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000165152.hg.1 |
ENST00000491735.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000165152.hg.1 |
NM_004508.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000165152.hg.1 |
ENST00000381344.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000165152.hg.1 |
ENST00000427898.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000165152.hg.1 |
ENST00000429642.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000165152.hg.1 |
ENST00000491735.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000165152.hg.1 |
NM_004508.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000165152.hg.1 |
TC1000009536.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000165150.hg.1 |
ENST00000381344.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000165150.hg.1 |
ENST00000491735.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000165150.hg.1 |
NM_004508.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000165150.hg.1 |
ENST00000381344.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000165150.hg.1 |
ENST00000491735.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000165150.hg.1 |
NM_004508.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000165150.hg.1 |
TC1000009536.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1000009536.hg.1 |
ENST00000381344.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009536.hg.1 |
ENST00000381344.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009536.hg.1 |
ENST00000381344.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009536.hg.1 |
ENST00000381344.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009536.hg.1 |
ENST00000381344.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009536.hg.1 |
ENST00000381344.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009536.hg.1 |
ENST00000381344.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009536.hg.1 |
ENST00000381344.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009536.hg.1 |
ENST00000381344.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009536.hg.1 |
ENST00000381344.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009536.hg.1 |
ENST00000381344.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009536.hg.1 |
ENST00000381344.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009536.hg.1 |
ENST00000427898.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009536.hg.1 |
ENST00000427898.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009536.hg.1 |
ENST00000427898.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009536.hg.1 |
ENST00000427898.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009536.hg.1 |
ENST00000427898.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009536.hg.1 |
ENST00000429642.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009536.hg.1 |
ENST00000429642.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009536.hg.1 |
ENST00000429642.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009536.hg.1 |
ENST00000429642.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009536.hg.1 |
ENST00000429642.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009536.hg.1 |
ENST00000429642.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009536.hg.1 |
ENST00000482091.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009536.hg.1 |
ENST00000482091.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009536.hg.1 |
ENST00000482091.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009536.hg.1 |
ENST00000482091.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009536.hg.1 |
ENST00000482091.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009536.hg.1 |
ENST00000491735.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009536.hg.1 |
ENST00000491735.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009536.hg.1 |
ENST00000491735.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009536.hg.1 |
ENST00000491735.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009536.hg.1 |
ENST00000491735.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009536.hg.1 |
ENST00000491735.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009536.hg.1 |
ENST00000491735.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009536.hg.1 |
ENST00000491735.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009536.hg.1 |
NM_004508.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009536.hg.1 |
NM_004508.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009536.hg.1 |
NM_004508.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009536.hg.1 |
NM_004508.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009536.hg.1 |
NM_004508.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009536.hg.1 |
NM_004508.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009536.hg.1 |
NM_004508.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009536.hg.1 |
NM_004508.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009536.hg.1 |
NM_004508.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009536.hg.1 |
NM_004508.3 |
Refseq |