Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PMEL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001200054.1 | NP_001186983.1 | 9 - 10 | 1808 | 92 | |
NM_001320121.1 | NP_001307050.1 | 10 - 11 | 1682 | 92 | ||
XM_011538685.1 | XP_011536987.1 | 10 - 11 | 1872 | 92 | ||
GenBank mRNA | M77348.1 | - | 9 - 10 | 1768 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Probe spans exons |