Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01008986.hg.1 |
TC01000553.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01008986.hg.1 |
ENST00000372343.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01008986.hg.1 |
ENST00000492152.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01008986.hg.1 |
NM_014652.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008079.hg.1 |
ENST00000372339.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008079.hg.1 |
ENST00000372339.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008079.hg.1 |
ENST00000372339.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008079.hg.1 |
ENST00000372339.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008079.hg.1 |
ENST00000372339.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008079.hg.1 |
ENST00000372339.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008079.hg.1 |
ENST00000372339.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008079.hg.1 |
ENST00000372343.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008079.hg.1 |
ENST00000372343.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008079.hg.1 |
ENST00000372343.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008079.hg.1 |
ENST00000372343.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008079.hg.1 |
ENST00000372343.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008079.hg.1 |
ENST00000372343.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008079.hg.1 |
ENST00000372343.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008079.hg.1 |
ENST00000372343.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008079.hg.1 |
ENST00000372343.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008079.hg.1 |
ENST00000372343.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008079.hg.1 |
ENST00000372343.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008079.hg.1 |
ENST00000372343.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008079.hg.1 |
ENST00000372343.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008079.hg.1 |
ENST00000372343.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008079.hg.1 |
ENST00000372343.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008079.hg.1 |
ENST00000372343.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008079.hg.1 |
ENST00000372343.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008079.hg.1 |
ENST00000372343.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008079.hg.1 |
ENST00000372343.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008079.hg.1 |
ENST00000372343.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008079.hg.1 |
ENST00000372343.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008079.hg.1 |
ENST00000372343.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008079.hg.1 |
ENST00000372343.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008079.hg.1 |
ENST00000372343.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008079.hg.1 |
ENST00000372343.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008079.hg.1 |
ENST00000372343.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008079.hg.1 |
ENST00000372343.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008079.hg.1 |
ENST00000372343.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008079.hg.1 |
ENST00000480902.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008079.hg.1 |
ENST00000480902.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008079.hg.1 |
ENST00000480902.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008079.hg.1 |
ENST00000486876.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008079.hg.1 |
ENST00000486876.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008079.hg.1 |
ENST00000486876.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008079.hg.1 |
ENST00000489061.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008079.hg.1 |
ENST00000489061.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008079.hg.1 |
ENST00000489061.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008079.hg.1 |
ENST00000489061.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008079.hg.1 |
ENST00000489773.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008079.hg.1 |
ENST00000489773.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008079.hg.1 |
ENST00000489773.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008079.hg.1 |
ENST00000489773.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008079.hg.1 |
ENST00000489773.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008079.hg.1 |
ENST00000489773.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008079.hg.1 |
ENST00000492152.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008079.hg.1 |
ENST00000492152.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008079.hg.1 |
ENST00000492152.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008079.hg.1 |
ENST00000492152.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008079.hg.1 |
ENST00000492152.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008079.hg.1 |
ENST00000492152.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008079.hg.1 |
ENST00000492152.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008079.hg.1 |
NM_014652.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008079.hg.1 |
NM_014652.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008079.hg.1 |
NM_014652.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008079.hg.1 |
NM_014652.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008079.hg.1 |
NM_014652.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008079.hg.1 |
NM_014652.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008079.hg.1 |
NM_014652.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008079.hg.1 |
NM_014652.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008079.hg.1 |
NM_014652.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008079.hg.1 |
NM_014652.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008079.hg.1 |
NM_014652.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008079.hg.1 |
NM_014652.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008079.hg.1 |
NM_014652.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008079.hg.1 |
NM_014652.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008079.hg.1 |
NM_014652.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008079.hg.1 |
NM_014652.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008079.hg.1 |
NM_014652.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008079.hg.1 |
NM_014652.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008079.hg.1 |
NM_014652.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008079.hg.1 |
NM_014652.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008079.hg.1 |
NM_014652.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008079.hg.1 |
NM_014652.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008079.hg.1 |
NM_014652.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008079.hg.1 |
NM_014652.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008079.hg.1 |
NM_014652.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008079.hg.1 |
NM_014652.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008079.hg.1 |
NM_014652.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008079.hg.1 |
NM_014652.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008079.hg.1 |
NM_014652.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01008987.hg.1 |
TC01000553.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01008987.hg.1 |
ENST00000372343.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01008987.hg.1 |
ENST00000492152.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01008987.hg.1 |
NM_014652.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100157264.hg.1 |
ENST00000372343.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100157264.hg.1 |
ENST00000489773.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100157264.hg.1 |
ENST00000492152.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100157264.hg.1 |
NM_014652.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100157264.hg.1 |
ENST00000372343.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100157264.hg.1 |
ENST00000489773.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100157264.hg.1 |
ENST00000492152.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100157264.hg.1 |
NM_014652.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100157264.hg.1 |
TC0100008079.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100008079.hg.1 |
ENST00000372339.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008079.hg.1 |
ENST00000372339.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008079.hg.1 |
ENST00000372339.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008079.hg.1 |
ENST00000372339.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008079.hg.1 |
ENST00000372339.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008079.hg.1 |
ENST00000372339.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008079.hg.1 |
ENST00000372339.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008079.hg.1 |
ENST00000372343.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008079.hg.1 |
ENST00000372343.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008079.hg.1 |
ENST00000372343.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008079.hg.1 |
ENST00000372343.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008079.hg.1 |
ENST00000372343.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008079.hg.1 |
ENST00000372343.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008079.hg.1 |
ENST00000372343.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008079.hg.1 |
ENST00000372343.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008079.hg.1 |
ENST00000372343.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008079.hg.1 |
ENST00000372343.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008079.hg.1 |
ENST00000372343.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008079.hg.1 |
ENST00000372343.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008079.hg.1 |
ENST00000372343.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008079.hg.1 |
ENST00000372343.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008079.hg.1 |
ENST00000372343.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008079.hg.1 |
ENST00000372343.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008079.hg.1 |
ENST00000372343.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008079.hg.1 |
ENST00000372343.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008079.hg.1 |
ENST00000372343.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008079.hg.1 |
ENST00000372343.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008079.hg.1 |
ENST00000372343.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008079.hg.1 |
ENST00000372343.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008079.hg.1 |
ENST00000372343.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008079.hg.1 |
ENST00000372343.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008079.hg.1 |
ENST00000372343.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008079.hg.1 |
ENST00000372343.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008079.hg.1 |
ENST00000372343.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008079.hg.1 |
ENST00000372343.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008079.hg.1 |
ENST00000480902.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008079.hg.1 |
ENST00000480902.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008079.hg.1 |
ENST00000480902.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008079.hg.1 |
ENST00000486876.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008079.hg.1 |
ENST00000486876.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008079.hg.1 |
ENST00000486876.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008079.hg.1 |
ENST00000489061.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008079.hg.1 |
ENST00000489061.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008079.hg.1 |
ENST00000489061.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008079.hg.1 |
ENST00000489061.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008079.hg.1 |
ENST00000489773.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008079.hg.1 |
ENST00000489773.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008079.hg.1 |
ENST00000489773.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008079.hg.1 |
ENST00000489773.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008079.hg.1 |
ENST00000489773.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008079.hg.1 |
ENST00000489773.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008079.hg.1 |
ENST00000492152.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008079.hg.1 |
ENST00000492152.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008079.hg.1 |
ENST00000492152.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008079.hg.1 |
ENST00000492152.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008079.hg.1 |
ENST00000492152.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008079.hg.1 |
ENST00000492152.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008079.hg.1 |
ENST00000492152.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008079.hg.1 |
NM_014652.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008079.hg.1 |
NM_014652.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008079.hg.1 |
NM_014652.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008079.hg.1 |
NM_014652.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008079.hg.1 |
NM_014652.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008079.hg.1 |
NM_014652.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008079.hg.1 |
NM_014652.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008079.hg.1 |
NM_014652.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008079.hg.1 |
NM_014652.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008079.hg.1 |
NM_014652.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008079.hg.1 |
NM_014652.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008079.hg.1 |
NM_014652.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008079.hg.1 |
NM_014652.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008079.hg.1 |
NM_014652.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008079.hg.1 |
NM_014652.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008079.hg.1 |
NM_014652.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008079.hg.1 |
NM_014652.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008079.hg.1 |
NM_014652.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008079.hg.1 |
NM_014652.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008079.hg.1 |
NM_014652.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008079.hg.1 |
NM_014652.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008079.hg.1 |
NM_014652.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008079.hg.1 |
NM_014652.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008079.hg.1 |
NM_014652.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008079.hg.1 |
NM_014652.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008079.hg.1 |
NM_014652.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008079.hg.1 |
NM_014652.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008079.hg.1 |
NM_014652.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008079.hg.1 |
NM_014652.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100157265.hg.1 |
ENST00000372343.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100157265.hg.1 |
ENST00000492152.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100157265.hg.1 |
NM_014652.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100157265.hg.1 |
ENST00000372343.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100157265.hg.1 |
ENST00000492152.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100157265.hg.1 |
NM_014652.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100157265.hg.1 |
TC0100008079.hg.1 |
Clariom D TC |