Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_004304.4 | NP_004295.2 | 23 - 24 | 4596 | 76 | |
GenBank mRNA | AB209477.4 | - | 23 - 24 | 4133 | 76 | |
EU788003.1 | - | 23 - 24 | 4551 | 76 | ||
EU788004.1 | - | 23 - 24 | 4551 | 76 | ||
EU788005.1 | - | 23 - 24 | 4551 | 76 | ||
EU788006.1 | - | 23 - 24 | 4551 | 76 | ||
EU788007.1 | - | 23 - 24 | 4551 | 76 | ||
EU788008.1 | - | 23 - 24 | 4551 | 76 | ||
EU788009.1 | - | 23 - 24 | 4551 | 76 | ||
EU788010.1 | - | 23 - 24 | 4551 | 76 | ||
FJ838786.1 | - | 23 - 24 | 4551 | 76 | ||
FJ838787.1 | - | 23 - 24 | 4551 | 76 | ||
GU128152.1 | - | 23 - 24 | 3644 | 76 | ||
GU128153.1 | - | 23 - 24 | 3644 | 76 | ||
GU128154.1 | - | 23 - 24 | 3644 | 76 | ||
GU128155.1 | - | 23 - 24 | 3644 | 76 | ||
GU128157.1 | - | 23 - 24 | 3644 | 76 | ||
LN864494.1 | - | 4 - 5 | 844 | 76 | ||
U62540.1 | - | 23 - 24 | 4555 | 76 | ||
U66559.1 | - | 23 - 24 | 3792 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Homo sapiens Inventoried Probe spans exons |