Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013314.hg.1 |
ENST00000270800.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013314.hg.1 |
ENST00000270800.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013314.hg.1 |
ENST00000270800.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013314.hg.1 |
ENST00000270800.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013314.hg.1 |
ENST00000270800.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013314.hg.1 |
ENST00000270800.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013314.hg.1 |
ENST00000270800.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013314.hg.1 |
ENST00000270800.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013314.hg.1 |
ENST00000270800.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013314.hg.1 |
ENST00000270800.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013314.hg.1 |
ENST00000270800.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013314.hg.1 |
NM_021258.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013314.hg.1 |
NM_021258.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013314.hg.1 |
NM_021258.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013314.hg.1 |
NM_021258.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013314.hg.1 |
NM_021258.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013314.hg.1 |
NM_021258.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013314.hg.1 |
NM_021258.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013314.hg.1 |
NM_021258.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013314.hg.1 |
NM_021258.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013314.hg.1 |
NM_021258.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013314.hg.1 |
NM_021258.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013314.hg.1 |
NM_021258.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01036444.hg.1 |
TC01002358.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01036444.hg.1 |
ENST00000270800.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01036444.hg.1 |
NM_021258.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013314.hg.1 |
ENST00000270800.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013314.hg.1 |
ENST00000270800.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013314.hg.1 |
ENST00000270800.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013314.hg.1 |
ENST00000270800.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013314.hg.1 |
ENST00000270800.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013314.hg.1 |
ENST00000270800.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013314.hg.1 |
ENST00000270800.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013314.hg.1 |
ENST00000270800.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013314.hg.1 |
ENST00000270800.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013314.hg.1 |
ENST00000270800.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013314.hg.1 |
ENST00000270800.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013314.hg.1 |
NM_021258.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013314.hg.1 |
NM_021258.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013314.hg.1 |
NM_021258.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013314.hg.1 |
NM_021258.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013314.hg.1 |
NM_021258.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013314.hg.1 |
NM_021258.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013314.hg.1 |
NM_021258.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013314.hg.1 |
NM_021258.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013314.hg.1 |
NM_021258.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013314.hg.1 |
NM_021258.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013314.hg.1 |
NM_021258.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013314.hg.1 |
NM_021258.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100198869.hg.1 |
ENST00000270800.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100198869.hg.1 |
NM_021258.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100198869.hg.1 |
ENST00000270800.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100198869.hg.1 |
NM_021258.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100198869.hg.1 |
TC0100013314.hg.1 |
Clariom D TC |