Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR07011664.hg.1 |
TC07000729.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07011664.hg.1 |
ENST00000265224.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07011664.hg.1 |
ENST00000357099.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07011664.hg.1 |
ENST00000417525.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07011664.hg.1 |
ENST00000433239.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07011664.hg.1 |
ENST00000477532.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07011664.hg.1 |
NM_019644.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008898.hg.1 |
ENST00000265224.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008898.hg.1 |
ENST00000265224.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008898.hg.1 |
ENST00000265224.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008898.hg.1 |
ENST00000265224.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008898.hg.1 |
ENST00000265224.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008898.hg.1 |
ENST00000265224.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008898.hg.1 |
ENST00000265224.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008898.hg.1 |
ENST00000265224.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008898.hg.1 |
ENST00000265224.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008898.hg.1 |
ENST00000265224.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008898.hg.1 |
ENST00000265224.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008898.hg.1 |
ENST00000265224.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008898.hg.1 |
ENST00000357099.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008898.hg.1 |
ENST00000357099.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008898.hg.1 |
ENST00000357099.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008898.hg.1 |
ENST00000357099.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008898.hg.1 |
ENST00000357099.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008898.hg.1 |
ENST00000357099.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008898.hg.1 |
ENST00000357099.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008898.hg.1 |
ENST00000357099.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008898.hg.1 |
ENST00000357099.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008898.hg.1 |
ENST00000357099.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008898.hg.1 |
ENST00000357099.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008898.hg.1 |
ENST00000357099.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008898.hg.1 |
ENST00000417525.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008898.hg.1 |
ENST00000417525.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008898.hg.1 |
ENST00000417525.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008898.hg.1 |
ENST00000417525.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008898.hg.1 |
ENST00000417525.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008898.hg.1 |
ENST00000417525.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008898.hg.1 |
ENST00000417525.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008898.hg.1 |
ENST00000417525.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008898.hg.1 |
ENST00000417525.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008898.hg.1 |
ENST00000417525.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008898.hg.1 |
ENST00000417525.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008898.hg.1 |
ENST00000433239.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008898.hg.1 |
ENST00000433239.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008898.hg.1 |
ENST00000433239.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008898.hg.1 |
ENST00000433239.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008898.hg.1 |
ENST00000433239.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008898.hg.1 |
ENST00000433239.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008898.hg.1 |
ENST00000433239.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008898.hg.1 |
ENST00000433239.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008898.hg.1 |
ENST00000433239.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008898.hg.1 |
ENST00000477532.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008898.hg.1 |
ENST00000477532.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008898.hg.1 |
ENST00000477532.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008898.hg.1 |
ENST00000477532.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008898.hg.1 |
ENST00000477532.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008898.hg.1 |
ENST00000477532.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008898.hg.1 |
ENST00000477532.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008898.hg.1 |
ENST00000477532.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008898.hg.1 |
ENST00000486422.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008898.hg.1 |
ENST00000486422.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008898.hg.1 |
ENST00000486422.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008898.hg.1 |
ENST00000486422.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008898.hg.1 |
ENST00000486422.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008898.hg.1 |
ENST00000490445.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008898.hg.1 |
ENST00000490445.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008898.hg.1 |
ENST00000490445.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008898.hg.1 |
ENST00000490445.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008898.hg.1 |
ENST00000622000.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008898.hg.1 |
ENST00000622000.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008898.hg.1 |
ENST00000622000.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008898.hg.1 |
ENST00000622000.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008898.hg.1 |
ENST00000622000.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008898.hg.1 |
ENST00000622000.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008898.hg.1 |
ENST00000622000.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008898.hg.1 |
ENST00000622000.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008898.hg.1 |
ENST00000622000.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008898.hg.1 |
NM_019644.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008898.hg.1 |
NM_019644.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008898.hg.1 |
NM_019644.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008898.hg.1 |
NM_019644.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008898.hg.1 |
NM_019644.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008898.hg.1 |
NM_019644.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008898.hg.1 |
NM_019644.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008898.hg.1 |
NM_019644.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008898.hg.1 |
NM_019644.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008898.hg.1 |
NM_019644.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008898.hg.1 |
NM_019644.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008898.hg.1 |
NM_019644.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07011665.hg.1 |
TC07000729.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07011665.hg.1 |
ENST00000265224.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07011665.hg.1 |
ENST00000357099.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07011665.hg.1 |
ENST00000417525.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07011665.hg.1 |
ENST00000433239.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07011665.hg.1 |
ENST00000477532.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07011665.hg.1 |
NM_019644.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700163122.hg.1 |
ENST00000265224.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700163122.hg.1 |
ENST00000357099.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700163122.hg.1 |
ENST00000417525.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700163122.hg.1 |
ENST00000433239.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700163122.hg.1 |
ENST00000477532.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700163122.hg.1 |
ENST00000622000.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700163122.hg.1 |
NM_019644.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700163122.hg.1 |
ENST00000265224.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700163122.hg.1 |
ENST00000357099.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700163122.hg.1 |
ENST00000417525.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700163122.hg.1 |
ENST00000433239.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700163122.hg.1 |
ENST00000477532.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700163122.hg.1 |
ENST00000622000.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700163122.hg.1 |
NM_019644.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700163122.hg.1 |
TC0700008898.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0700008898.hg.1 |
ENST00000265224.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008898.hg.1 |
ENST00000265224.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008898.hg.1 |
ENST00000265224.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008898.hg.1 |
ENST00000265224.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008898.hg.1 |
ENST00000265224.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008898.hg.1 |
ENST00000265224.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008898.hg.1 |
ENST00000265224.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008898.hg.1 |
ENST00000265224.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008898.hg.1 |
ENST00000265224.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008898.hg.1 |
ENST00000265224.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008898.hg.1 |
ENST00000265224.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008898.hg.1 |
ENST00000265224.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008898.hg.1 |
ENST00000357099.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008898.hg.1 |
ENST00000357099.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008898.hg.1 |
ENST00000357099.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008898.hg.1 |
ENST00000357099.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008898.hg.1 |
ENST00000357099.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008898.hg.1 |
ENST00000357099.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008898.hg.1 |
ENST00000357099.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008898.hg.1 |
ENST00000357099.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008898.hg.1 |
ENST00000357099.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008898.hg.1 |
ENST00000357099.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008898.hg.1 |
ENST00000357099.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008898.hg.1 |
ENST00000357099.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008898.hg.1 |
ENST00000417525.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008898.hg.1 |
ENST00000417525.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008898.hg.1 |
ENST00000417525.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008898.hg.1 |
ENST00000417525.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008898.hg.1 |
ENST00000417525.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008898.hg.1 |
ENST00000417525.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008898.hg.1 |
ENST00000417525.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008898.hg.1 |
ENST00000417525.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008898.hg.1 |
ENST00000417525.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008898.hg.1 |
ENST00000417525.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008898.hg.1 |
ENST00000417525.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008898.hg.1 |
ENST00000433239.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008898.hg.1 |
ENST00000433239.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008898.hg.1 |
ENST00000433239.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008898.hg.1 |
ENST00000433239.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008898.hg.1 |
ENST00000433239.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008898.hg.1 |
ENST00000433239.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008898.hg.1 |
ENST00000433239.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008898.hg.1 |
ENST00000433239.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008898.hg.1 |
ENST00000433239.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008898.hg.1 |
ENST00000477532.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008898.hg.1 |
ENST00000477532.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008898.hg.1 |
ENST00000477532.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008898.hg.1 |
ENST00000477532.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008898.hg.1 |
ENST00000477532.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008898.hg.1 |
ENST00000477532.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008898.hg.1 |
ENST00000477532.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008898.hg.1 |
ENST00000477532.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008898.hg.1 |
ENST00000486422.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008898.hg.1 |
ENST00000486422.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008898.hg.1 |
ENST00000486422.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008898.hg.1 |
ENST00000486422.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008898.hg.1 |
ENST00000486422.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008898.hg.1 |
ENST00000490445.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008898.hg.1 |
ENST00000490445.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008898.hg.1 |
ENST00000490445.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008898.hg.1 |
ENST00000490445.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008898.hg.1 |
ENST00000622000.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008898.hg.1 |
ENST00000622000.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008898.hg.1 |
ENST00000622000.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008898.hg.1 |
ENST00000622000.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008898.hg.1 |
ENST00000622000.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008898.hg.1 |
ENST00000622000.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008898.hg.1 |
ENST00000622000.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008898.hg.1 |
ENST00000622000.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008898.hg.1 |
ENST00000622000.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008898.hg.1 |
NM_019644.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008898.hg.1 |
NM_019644.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008898.hg.1 |
NM_019644.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008898.hg.1 |
NM_019644.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008898.hg.1 |
NM_019644.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008898.hg.1 |
NM_019644.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008898.hg.1 |
NM_019644.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008898.hg.1 |
NM_019644.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008898.hg.1 |
NM_019644.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008898.hg.1 |
NM_019644.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008898.hg.1 |
NM_019644.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008898.hg.1 |
NM_019644.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700163124.hg.1 |
ENST00000265224.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700163124.hg.1 |
ENST00000357099.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700163124.hg.1 |
ENST00000417525.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700163124.hg.1 |
ENST00000433239.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700163124.hg.1 |
ENST00000477532.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700163124.hg.1 |
ENST00000622000.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700163124.hg.1 |
NM_019644.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700163124.hg.1 |
ENST00000265224.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700163124.hg.1 |
ENST00000357099.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700163124.hg.1 |
ENST00000417525.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700163124.hg.1 |
ENST00000433239.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700163124.hg.1 |
ENST00000477532.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700163124.hg.1 |
ENST00000622000.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700163124.hg.1 |
NM_019644.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700163124.hg.1 |
TC0700008898.hg.1 |
Clariom D TC |