Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011389.hg.1 |
ENST00000367096.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011389.hg.1 |
ENST00000367096.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011389.hg.1 |
ENST00000367096.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011389.hg.1 |
ENST00000367096.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011389.hg.1 |
ENST00000367096.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011389.hg.1 |
ENST00000367096.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011389.hg.1 |
ENST00000367096.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011389.hg.1 |
ENST00000367096.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011389.hg.1 |
ENST00000367096.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011389.hg.1 |
ENST00000367098.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011389.hg.1 |
ENST00000367098.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011389.hg.1 |
ENST00000367098.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011389.hg.1 |
ENST00000367098.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011389.hg.1 |
ENST00000367098.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011389.hg.1 |
ENST00000367098.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011389.hg.1 |
ENST00000367098.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011389.hg.1 |
ENST00000367098.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011389.hg.1 |
ENST00000367098.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011389.hg.1 |
ENST00000367098.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011389.hg.1 |
ENST00000367098.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011389.hg.1 |
ENST00000391930.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011389.hg.1 |
ENST00000391930.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011389.hg.1 |
ENST00000391930.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011389.hg.1 |
ENST00000391930.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011389.hg.1 |
ENST00000391930.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011389.hg.1 |
ENST00000391930.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011389.hg.1 |
ENST00000391930.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011389.hg.1 |
ENST00000391930.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011389.hg.1 |
ENST00000391930.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011389.hg.1 |
NM_018724.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011389.hg.1 |
NM_018724.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011389.hg.1 |
NM_018724.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011389.hg.1 |
NM_018724.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011389.hg.1 |
NM_018724.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011389.hg.1 |
NM_018724.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011389.hg.1 |
NM_018724.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011389.hg.1 |
NM_018724.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011389.hg.1 |
NM_018724.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011389.hg.1 |
NM_018724.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01026981.hg.1 |
TC01001729.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01026981.hg.1 |
ENST00000367096.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01026981.hg.1 |
ENST00000367098.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01026981.hg.1 |
NM_018724.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011389.hg.1 |
ENST00000367096.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011389.hg.1 |
ENST00000367096.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011389.hg.1 |
ENST00000367096.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011389.hg.1 |
ENST00000367096.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011389.hg.1 |
ENST00000367096.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011389.hg.1 |
ENST00000367096.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011389.hg.1 |
ENST00000367096.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011389.hg.1 |
ENST00000367096.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011389.hg.1 |
ENST00000367096.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011389.hg.1 |
ENST00000367098.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011389.hg.1 |
ENST00000367098.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011389.hg.1 |
ENST00000367098.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011389.hg.1 |
ENST00000367098.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011389.hg.1 |
ENST00000367098.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011389.hg.1 |
ENST00000367098.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011389.hg.1 |
ENST00000367098.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011389.hg.1 |
ENST00000367098.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011389.hg.1 |
ENST00000367098.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011389.hg.1 |
ENST00000367098.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011389.hg.1 |
ENST00000367098.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011389.hg.1 |
ENST00000391930.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011389.hg.1 |
ENST00000391930.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011389.hg.1 |
ENST00000391930.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011389.hg.1 |
ENST00000391930.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011389.hg.1 |
ENST00000391930.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011389.hg.1 |
ENST00000391930.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011389.hg.1 |
ENST00000391930.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011389.hg.1 |
ENST00000391930.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011389.hg.1 |
ENST00000391930.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011389.hg.1 |
NM_018724.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011389.hg.1 |
NM_018724.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011389.hg.1 |
NM_018724.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011389.hg.1 |
NM_018724.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011389.hg.1 |
NM_018724.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011389.hg.1 |
NM_018724.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011389.hg.1 |
NM_018724.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011389.hg.1 |
NM_018724.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011389.hg.1 |
NM_018724.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011389.hg.1 |
NM_018724.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100183745.hg.1 |
ENST00000367096.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100183745.hg.1 |
ENST00000367098.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100183745.hg.1 |
NM_018724.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100183745.hg.1 |
ENST00000367096.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100183745.hg.1 |
ENST00000367098.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100183745.hg.1 |
NM_018724.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100183745.hg.1 |
TC0100011389.hg.1 |
Clariom D TC |