Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP12A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001185085.1 | NP_001172014.1 | 20 - 21 | 3244 | 66 | |
NM_001676.5 | NP_001667.4 | 20 - 21 | 3226 | 66 | ||
GenBank mRNA | AK292968.1 | - | 20 - 21 | 3226 | 66 | |
AK303498.1 | - | 13 - 14 | 1978 | 66 | ||
BC031609.1 | - | 20 - 21 | 3078 | 5176407 | 66 | |
CA450333.1 | - | 3 - 4 | 66 | |||
U02076.1 | - | 20 - 21 | 3060 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Transporter Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |