Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012002.hg.1 |
ENST00000371688.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012002.hg.1 |
ENST00000371688.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012002.hg.1 |
ENST00000371688.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012002.hg.1 |
ENST00000371688.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012002.hg.1 |
ENST00000371688.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012002.hg.1 |
ENST00000371688.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012002.hg.1 |
ENST00000371688.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012002.hg.1 |
ENST00000371688.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012002.hg.1 |
ENST00000479767.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012002.hg.1 |
ENST00000479767.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012002.hg.1 |
ENST00000479767.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012002.hg.1 |
ENST00000479767.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012002.hg.1 |
ENST00000479767.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012002.hg.1 |
ENST00000479767.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012002.hg.1 |
ENST00000479767.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012002.hg.1 |
ENST00000479767.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012002.hg.1 |
ENST00000479767.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012002.hg.1 |
ENST00000480597.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012002.hg.1 |
ENST00000480597.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012002.hg.1 |
ENST00000480597.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012002.hg.1 |
ENST00000480597.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012002.hg.1 |
ENST00000480597.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012002.hg.1 |
ENST00000480597.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012002.hg.1 |
ENST00000480597.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012002.hg.1 |
ENST00000482893.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012002.hg.1 |
ENST00000482893.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012002.hg.1 |
ENST00000482893.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012002.hg.1 |
ENST00000482893.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012002.hg.1 |
ENST00000482893.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012002.hg.1 |
ENST00000482893.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012002.hg.1 |
ENST00000482893.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012002.hg.1 |
ENST00000482893.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012002.hg.1 |
ENST00000482893.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012002.hg.1 |
ENST00000482893.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012002.hg.1 |
ENST00000482893.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012002.hg.1 |
ENST00000482893.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012002.hg.1 |
ENST00000482893.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012002.hg.1 |
ENST00000612714.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012002.hg.1 |
ENST00000612714.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012002.hg.1 |
ENST00000612714.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012002.hg.1 |
ENST00000612714.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012002.hg.1 |
ENST00000612714.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012002.hg.1 |
ENST00000612714.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012002.hg.1 |
ENST00000612714.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012002.hg.1 |
ENST00000612714.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012002.hg.1 |
NM_178469.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012002.hg.1 |
NM_178469.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012002.hg.1 |
NM_178469.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012002.hg.1 |
NM_178469.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012002.hg.1 |
NM_178469.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012002.hg.1 |
NM_178469.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012002.hg.1 |
NM_178469.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012002.hg.1 |
NM_178469.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012002.hg.1 |
NM_178469.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09022402.hg.1 |
TC09001741.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09022402.hg.1 |
ENST00000371688.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09022402.hg.1 |
ENST00000482893.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09022402.hg.1 |
NM_178469.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012002.hg.1 |
ENST00000371688.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012002.hg.1 |
ENST00000371688.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012002.hg.1 |
ENST00000371688.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012002.hg.1 |
ENST00000371688.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012002.hg.1 |
ENST00000371688.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012002.hg.1 |
ENST00000371688.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012002.hg.1 |
ENST00000371688.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012002.hg.1 |
ENST00000371688.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012002.hg.1 |
ENST00000479767.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012002.hg.1 |
ENST00000479767.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012002.hg.1 |
ENST00000479767.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012002.hg.1 |
ENST00000479767.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012002.hg.1 |
ENST00000479767.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012002.hg.1 |
ENST00000479767.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012002.hg.1 |
ENST00000479767.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012002.hg.1 |
ENST00000479767.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012002.hg.1 |
ENST00000479767.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012002.hg.1 |
ENST00000480597.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012002.hg.1 |
ENST00000480597.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012002.hg.1 |
ENST00000480597.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012002.hg.1 |
ENST00000480597.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012002.hg.1 |
ENST00000480597.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012002.hg.1 |
ENST00000480597.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012002.hg.1 |
ENST00000480597.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012002.hg.1 |
ENST00000482893.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012002.hg.1 |
ENST00000482893.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012002.hg.1 |
ENST00000482893.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012002.hg.1 |
ENST00000482893.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012002.hg.1 |
ENST00000482893.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012002.hg.1 |
ENST00000482893.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012002.hg.1 |
ENST00000482893.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012002.hg.1 |
ENST00000482893.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012002.hg.1 |
ENST00000482893.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012002.hg.1 |
ENST00000482893.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012002.hg.1 |
ENST00000482893.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012002.hg.1 |
ENST00000482893.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012002.hg.1 |
ENST00000482893.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012002.hg.1 |
ENST00000612714.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012002.hg.1 |
ENST00000612714.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012002.hg.1 |
ENST00000612714.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012002.hg.1 |
ENST00000612714.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012002.hg.1 |
ENST00000612714.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012002.hg.1 |
ENST00000612714.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012002.hg.1 |
ENST00000612714.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012002.hg.1 |
ENST00000612714.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012002.hg.1 |
NM_178469.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012002.hg.1 |
NM_178469.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012002.hg.1 |
NM_178469.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012002.hg.1 |
NM_178469.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012002.hg.1 |
NM_178469.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012002.hg.1 |
NM_178469.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012002.hg.1 |
NM_178469.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012002.hg.1 |
NM_178469.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012002.hg.1 |
NM_178469.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900182368.hg.1 |
ENST00000371688.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900182368.hg.1 |
ENST00000479767.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900182368.hg.1 |
ENST00000480597.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900182368.hg.1 |
ENST00000482893.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900182368.hg.1 |
ENST00000612714.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900182368.hg.1 |
NM_178469.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900182368.hg.1 |
ENST00000371688.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900182368.hg.1 |
ENST00000479767.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900182368.hg.1 |
ENST00000480597.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900182368.hg.1 |
ENST00000482893.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900182368.hg.1 |
ENST00000612714.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900182368.hg.1 |
NM_178469.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900182368.hg.1 |
TC0900012002.hg.1 |
Clariom D TC |