Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR08005427.hg.1 |
TC08000382.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08005427.hg.1 |
ENST00000260129.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08005427.hg.1 |
NM_024831.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007681.hg.1 |
ENST00000260129.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007681.hg.1 |
ENST00000260129.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007681.hg.1 |
ENST00000260129.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007681.hg.1 |
ENST00000260129.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007681.hg.1 |
ENST00000260129.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007681.hg.1 |
ENST00000260129.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007681.hg.1 |
ENST00000260129.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007681.hg.1 |
ENST00000260129.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007681.hg.1 |
ENST00000260129.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007681.hg.1 |
ENST00000260129.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007681.hg.1 |
ENST00000260129.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007681.hg.1 |
ENST00000260129.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007681.hg.1 |
ENST00000260129.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007681.hg.1 |
ENST00000260129.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007681.hg.1 |
ENST00000260129.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007681.hg.1 |
ENST00000260129.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007681.hg.1 |
ENST00000260129.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007681.hg.1 |
ENST00000260129.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007681.hg.1 |
ENST00000519494.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007681.hg.1 |
ENST00000519494.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007681.hg.1 |
ENST00000519494.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007681.hg.1 |
ENST00000519494.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007681.hg.1 |
ENST00000519494.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007681.hg.1 |
ENST00000523948.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007681.hg.1 |
ENST00000523948.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007681.hg.1 |
ENST00000523948.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007681.hg.1 |
ENST00000523948.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007681.hg.1 |
ENST00000523948.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007681.hg.1 |
ENST00000523948.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007681.hg.1 |
ENST00000523948.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007681.hg.1 |
ENST00000523948.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007681.hg.1 |
ENST00000523948.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007681.hg.1 |
ENST00000523948.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007681.hg.1 |
ENST00000523948.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007681.hg.1 |
ENST00000523948.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007681.hg.1 |
ENST00000523948.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007681.hg.1 |
ENST00000523948.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007681.hg.1 |
NM_024831.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007681.hg.1 |
NM_024831.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007681.hg.1 |
NM_024831.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007681.hg.1 |
NM_024831.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007681.hg.1 |
NM_024831.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007681.hg.1 |
NM_024831.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007681.hg.1 |
NM_024831.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007681.hg.1 |
NM_024831.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007681.hg.1 |
NM_024831.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007681.hg.1 |
NM_024831.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007681.hg.1 |
NM_024831.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007681.hg.1 |
NM_024831.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007681.hg.1 |
NM_024831.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007681.hg.1 |
NM_024831.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007681.hg.1 |
NM_024831.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007681.hg.1 |
NM_024831.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007681.hg.1 |
NM_024831.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007681.hg.1 |
ENST00000260129.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007681.hg.1 |
ENST00000260129.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007681.hg.1 |
ENST00000260129.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007681.hg.1 |
ENST00000260129.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007681.hg.1 |
ENST00000260129.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007681.hg.1 |
ENST00000260129.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007681.hg.1 |
ENST00000260129.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007681.hg.1 |
ENST00000260129.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007681.hg.1 |
ENST00000260129.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007681.hg.1 |
ENST00000260129.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007681.hg.1 |
ENST00000260129.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007681.hg.1 |
ENST00000260129.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007681.hg.1 |
ENST00000260129.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007681.hg.1 |
ENST00000260129.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007681.hg.1 |
ENST00000260129.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007681.hg.1 |
ENST00000260129.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007681.hg.1 |
ENST00000260129.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007681.hg.1 |
ENST00000260129.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007681.hg.1 |
ENST00000519494.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007681.hg.1 |
ENST00000519494.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007681.hg.1 |
ENST00000519494.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007681.hg.1 |
ENST00000519494.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007681.hg.1 |
ENST00000519494.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007681.hg.1 |
ENST00000523948.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007681.hg.1 |
ENST00000523948.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007681.hg.1 |
ENST00000523948.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007681.hg.1 |
ENST00000523948.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007681.hg.1 |
ENST00000523948.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007681.hg.1 |
ENST00000523948.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007681.hg.1 |
ENST00000523948.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007681.hg.1 |
ENST00000523948.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007681.hg.1 |
ENST00000523948.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007681.hg.1 |
ENST00000523948.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007681.hg.1 |
ENST00000523948.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007681.hg.1 |
ENST00000523948.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007681.hg.1 |
ENST00000523948.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007681.hg.1 |
ENST00000523948.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007681.hg.1 |
NM_024831.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007681.hg.1 |
NM_024831.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007681.hg.1 |
NM_024831.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007681.hg.1 |
NM_024831.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007681.hg.1 |
NM_024831.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007681.hg.1 |
NM_024831.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007681.hg.1 |
NM_024831.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007681.hg.1 |
NM_024831.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007681.hg.1 |
NM_024831.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007681.hg.1 |
NM_024831.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007681.hg.1 |
NM_024831.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007681.hg.1 |
NM_024831.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007681.hg.1 |
NM_024831.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007681.hg.1 |
NM_024831.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007681.hg.1 |
NM_024831.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007681.hg.1 |
NM_024831.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007681.hg.1 |
NM_024831.7 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800152816.hg.1 |
ENST00000260129.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800152816.hg.1 |
NM_024831.7 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800152816.hg.1 |
ENST00000260129.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800152816.hg.1 |
NM_024831.7 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800152816.hg.1 |
TC0800007681.hg.1 |
Clariom D TC |