Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01024300.hg.1 |
TC01001572.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01024300.hg.1 |
ENST00000357570.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01024300.hg.1 |
ENST00000358338.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01024300.hg.1 |
ENST00000360108.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01024300.hg.1 |
ENST00000367567.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01024300.hg.1 |
ENST00000367570.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01024300.hg.1 |
ENST00000367573.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01024300.hg.1 |
ENST00000526775.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01024300.hg.1 |
NM_000721.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01024300.hg.1 |
NM_001205293.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01024300.hg.1 |
NM_001205294.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100179577.hg.1 |
ENST00000357570.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100179577.hg.1 |
ENST00000358338.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100179577.hg.1 |
ENST00000360108.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100179577.hg.1 |
ENST00000367567.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100179577.hg.1 |
ENST00000367570.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100179577.hg.1 |
ENST00000367573.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100179577.hg.1 |
ENST00000621551.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100179577.hg.1 |
ENST00000621791.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100179577.hg.1 |
NM_000721.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100179577.hg.1 |
NM_001205293.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100179577.hg.1 |
NM_001205294.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100179577.hg.1 |
ENST00000357570.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100179577.hg.1 |
ENST00000358338.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100179577.hg.1 |
ENST00000360108.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100179577.hg.1 |
ENST00000367567.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100179577.hg.1 |
ENST00000367570.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100179577.hg.1 |
ENST00000367573.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100179577.hg.1 |
ENST00000621551.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100179577.hg.1 |
ENST00000621791.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100179577.hg.1 |
NM_000721.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100179577.hg.1 |
NM_001205293.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100179577.hg.1 |
NM_001205294.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100179577.hg.1 |
TC0100010824.hg.1 |
Clariom D TC |