Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013600.hg.1 |
ENST00000397899.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013600.hg.1 |
ENST00000397899.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013600.hg.1 |
ENST00000397899.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013600.hg.1 |
ENST00000397899.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013600.hg.1 |
ENST00000397899.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013600.hg.1 |
ENST00000397899.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013600.hg.1 |
ENST00000397899.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013600.hg.1 |
ENST00000397899.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013600.hg.1 |
ENST00000397899.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013600.hg.1 |
ENST00000397899.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013600.hg.1 |
ENST00000397899.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013600.hg.1 |
ENST00000397899.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013600.hg.1 |
ENST00000397899.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013600.hg.1 |
ENST00000397899.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013600.hg.1 |
ENST00000397899.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013600.hg.1 |
ENST00000415261.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013600.hg.1 |
ENST00000415261.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013600.hg.1 |
ENST00000415261.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013600.hg.1 |
ENST00000415261.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013600.hg.1 |
ENST00000415261.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013600.hg.1 |
ENST00000415261.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013600.hg.1 |
ENST00000420294.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013600.hg.1 |
ENST00000420294.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013600.hg.1 |
ENST00000420294.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013600.hg.1 |
ENST00000420294.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013600.hg.1 |
ENST00000420294.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013600.hg.1 |
ENST00000420294.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013600.hg.1 |
ENST00000423771.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013600.hg.1 |
ENST00000423771.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013600.hg.1 |
ENST00000423771.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013600.hg.1 |
ENST00000423771.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013600.hg.1 |
ENST00000423771.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013600.hg.1 |
ENST00000423771.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013600.hg.1 |
ENST00000423771.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013600.hg.1 |
ENST00000428096.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013600.hg.1 |
ENST00000428096.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013600.hg.1 |
ENST00000428096.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013600.hg.1 |
ENST00000428096.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013600.hg.1 |
ENST00000428096.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013600.hg.1 |
ENST00000428096.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013600.hg.1 |
ENST00000428096.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013600.hg.1 |
ENST00000428096.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013600.hg.1 |
ENST00000462314.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013600.hg.1 |
ENST00000462314.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013600.hg.1 |
ENST00000462314.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013600.hg.1 |
ENST00000462314.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013600.hg.1 |
ENST00000462314.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013600.hg.1 |
ENST00000462314.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013600.hg.1 |
ENST00000464413.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013600.hg.1 |
ENST00000464413.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013600.hg.1 |
ENST00000612523.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013600.hg.1 |
ENST00000612523.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013600.hg.1 |
ENST00000612523.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013600.hg.1 |
ENST00000612523.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013600.hg.1 |
ENST00000612523.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013600.hg.1 |
ENST00000612523.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013600.hg.1 |
ENST00000612523.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013600.hg.1 |
ENST00000612523.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013600.hg.1 |
ENST00000612523.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013600.hg.1 |
ENST00000612523.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013600.hg.1 |
ENST00000612523.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013600.hg.1 |
ENST00000612523.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013600.hg.1 |
ENST00000612523.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013600.hg.1 |
ENST00000612523.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013600.hg.1 |
NM_207362.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013600.hg.1 |
NM_207362.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013600.hg.1 |
NM_207362.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013600.hg.1 |
NM_207362.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013600.hg.1 |
NM_207362.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013600.hg.1 |
NM_207362.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013600.hg.1 |
NM_207362.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013600.hg.1 |
NM_207362.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013600.hg.1 |
NM_207362.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013600.hg.1 |
NM_207362.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013600.hg.1 |
NM_207362.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013600.hg.1 |
NM_207362.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013600.hg.1 |
NM_207362.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013600.hg.1 |
NM_207362.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02034617.hg.1 |
TC02002129.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02034617.hg.1 |
ENST00000397899.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02034617.hg.1 |
NM_207362.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02034619.hg.1 |
TC02002129.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02034619.hg.1 |
ENST00000397899.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02034619.hg.1 |
NM_207362.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013600.hg.1 |
ENST00000397899.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013600.hg.1 |
ENST00000397899.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013600.hg.1 |
ENST00000397899.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013600.hg.1 |
ENST00000397899.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013600.hg.1 |
ENST00000397899.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013600.hg.1 |
ENST00000397899.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013600.hg.1 |
ENST00000397899.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013600.hg.1 |
ENST00000397899.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013600.hg.1 |
ENST00000397899.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013600.hg.1 |
ENST00000397899.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013600.hg.1 |
ENST00000397899.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013600.hg.1 |
ENST00000397899.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013600.hg.1 |
ENST00000397899.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013600.hg.1 |
ENST00000397899.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013600.hg.1 |
ENST00000397899.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013600.hg.1 |
ENST00000415261.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013600.hg.1 |
ENST00000415261.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013600.hg.1 |
ENST00000415261.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013600.hg.1 |
ENST00000415261.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013600.hg.1 |
ENST00000415261.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013600.hg.1 |
ENST00000415261.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013600.hg.1 |
ENST00000420294.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013600.hg.1 |
ENST00000420294.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013600.hg.1 |
ENST00000420294.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013600.hg.1 |
ENST00000420294.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013600.hg.1 |
ENST00000420294.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013600.hg.1 |
ENST00000420294.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013600.hg.1 |
ENST00000423771.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013600.hg.1 |
ENST00000423771.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013600.hg.1 |
ENST00000423771.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013600.hg.1 |
ENST00000423771.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013600.hg.1 |
ENST00000423771.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013600.hg.1 |
ENST00000423771.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013600.hg.1 |
ENST00000423771.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013600.hg.1 |
ENST00000428096.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013600.hg.1 |
ENST00000428096.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013600.hg.1 |
ENST00000428096.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013600.hg.1 |
ENST00000428096.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013600.hg.1 |
ENST00000428096.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013600.hg.1 |
ENST00000428096.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013600.hg.1 |
ENST00000428096.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013600.hg.1 |
ENST00000428096.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013600.hg.1 |
ENST00000462314.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013600.hg.1 |
ENST00000462314.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013600.hg.1 |
ENST00000462314.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013600.hg.1 |
ENST00000462314.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013600.hg.1 |
ENST00000462314.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013600.hg.1 |
ENST00000462314.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013600.hg.1 |
ENST00000464413.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013600.hg.1 |
ENST00000464413.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013600.hg.1 |
ENST00000612523.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013600.hg.1 |
ENST00000612523.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013600.hg.1 |
ENST00000612523.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013600.hg.1 |
ENST00000612523.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013600.hg.1 |
ENST00000612523.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013600.hg.1 |
ENST00000612523.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013600.hg.1 |
ENST00000612523.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013600.hg.1 |
ENST00000612523.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013600.hg.1 |
ENST00000612523.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013600.hg.1 |
ENST00000612523.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013600.hg.1 |
ENST00000612523.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013600.hg.1 |
ENST00000612523.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013600.hg.1 |
ENST00000612523.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013600.hg.1 |
ENST00000612523.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013600.hg.1 |
NM_207362.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013600.hg.1 |
NM_207362.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013600.hg.1 |
NM_207362.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013600.hg.1 |
NM_207362.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013600.hg.1 |
NM_207362.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013600.hg.1 |
NM_207362.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013600.hg.1 |
NM_207362.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013600.hg.1 |
NM_207362.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013600.hg.1 |
NM_207362.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013600.hg.1 |
NM_207362.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013600.hg.1 |
NM_207362.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013600.hg.1 |
NM_207362.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013600.hg.1 |
NM_207362.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013600.hg.1 |
NM_207362.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200199696.hg.1 |
ENST00000397899.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200199696.hg.1 |
ENST00000612523.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200199696.hg.1 |
NM_207362.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200199696.hg.1 |
ENST00000397899.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200199696.hg.1 |
ENST00000612523.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200199696.hg.1 |
NM_207362.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200199696.hg.1 |
TC0200013600.hg.1 |
Clariom D TC |