Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KMT2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001197104.1 | NP_001184033.1 | 18 - 19 | 5385 | 111 | |
NM_005933.3 | NP_005924.2 | 18 - 19 | 5376 | 111 | ||
XM_006718839.3 | XP_006718902.2 | 18 - 19 | 2860 | 111 | ||
XM_011542829.2 | XP_011541131.1 | 19 - 20 | 5476 | 111 | ||
XM_011542830.2 | XP_011541132.1 | 19 - 20 | 5473 | 111 | ||
XM_011542831.2 | XP_011541133.1 | 19 - 20 | 5467 | 111 | ||
XM_011542833.2 | XP_011541135.1 | 19 - 20 | 2959 | 111 | ||
GenBank mRNA | AB209508.1 | - | 16 - 17 | 2920 | 111 | |
D14540.1 | - | 18 - 19 | 5376 | 111 | ||
L01986.1 | - | 12 - 13 | 1407 | 111 | ||
L04284.1 | - | 18 - 19 | 5353 | 111 | ||
L04731.1 | - | 18 - 19 | 5176 | 111 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons |