Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09013601.hg.1 |
TC09001038.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09013601.hg.1 |
ENST00000351266.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09013601.hg.1 |
ENST00000378980.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09013601.hg.1 |
NM_001207011.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09013601.hg.1 |
NM_001842.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09013601.hg.1 |
NM_147164.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009905.hg.1 |
ENST00000351266.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009905.hg.1 |
ENST00000351266.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009905.hg.1 |
ENST00000351266.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009905.hg.1 |
ENST00000351266.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009905.hg.1 |
ENST00000351266.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009905.hg.1 |
ENST00000351266.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009905.hg.1 |
ENST00000351266.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009905.hg.1 |
ENST00000351266.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009905.hg.1 |
ENST00000351266.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009905.hg.1 |
ENST00000378980.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009905.hg.1 |
ENST00000378980.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009905.hg.1 |
ENST00000378980.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009905.hg.1 |
ENST00000378980.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009905.hg.1 |
ENST00000378980.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009905.hg.1 |
ENST00000378980.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009905.hg.1 |
ENST00000378980.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009905.hg.1 |
ENST00000378980.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009905.hg.1 |
ENST00000378980.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009905.hg.1 |
ENST00000378980.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009905.hg.1 |
ENST00000378980.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009905.hg.1 |
ENST00000417345.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009905.hg.1 |
ENST00000417345.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009905.hg.1 |
ENST00000417345.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009905.hg.1 |
ENST00000417345.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009905.hg.1 |
ENST00000417345.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009905.hg.1 |
ENST00000417345.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009905.hg.1 |
ENST00000417345.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009905.hg.1 |
ENST00000610543.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009905.hg.1 |
ENST00000610543.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009905.hg.1 |
ENST00000610543.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009905.hg.1 |
ENST00000610543.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009905.hg.1 |
ENST00000610543.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009905.hg.1 |
ENST00000610543.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009905.hg.1 |
ENST00000610543.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009905.hg.1 |
ENST00000610543.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009905.hg.1 |
ENST00000610543.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009905.hg.1 |
ENST00000610543.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009905.hg.1 |
NM_001207011.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009905.hg.1 |
NM_001207011.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009905.hg.1 |
NM_001207011.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009905.hg.1 |
NM_001207011.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009905.hg.1 |
NM_001207011.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009905.hg.1 |
NM_001207011.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009905.hg.1 |
NM_001207011.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009905.hg.1 |
NM_001207011.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009905.hg.1 |
NM_001207011.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009905.hg.1 |
NM_001207011.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009905.hg.1 |
NM_001842.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009905.hg.1 |
NM_001842.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009905.hg.1 |
NM_001842.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009905.hg.1 |
NM_001842.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009905.hg.1 |
NM_001842.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009905.hg.1 |
NM_001842.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009905.hg.1 |
NM_001842.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009905.hg.1 |
NM_001842.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009905.hg.1 |
NM_001842.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009905.hg.1 |
NM_001842.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009905.hg.1 |
NM_147164.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009905.hg.1 |
NM_147164.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009905.hg.1 |
NM_147164.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009905.hg.1 |
NM_147164.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009905.hg.1 |
NM_147164.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009905.hg.1 |
NM_147164.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009905.hg.1 |
NM_147164.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009905.hg.1 |
NM_147164.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009905.hg.1 |
NM_147164.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009905.hg.1 |
NM_147164.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009905.hg.1 |
NM_147164.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900167873.hg.1 |
ENST00000351266.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900167873.hg.1 |
ENST00000378980.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900167873.hg.1 |
ENST00000417345.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900167873.hg.1 |
ENST00000610543.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900167873.hg.1 |
NM_001207011.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900167873.hg.1 |
NM_001842.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900167873.hg.1 |
NM_147164.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900167873.hg.1 |
ENST00000351266.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900167873.hg.1 |
ENST00000378980.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900167873.hg.1 |
ENST00000417345.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900167873.hg.1 |
ENST00000610543.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900167873.hg.1 |
NM_001207011.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900167873.hg.1 |
NM_001842.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900167873.hg.1 |
NM_147164.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900167873.hg.1 |
TC0900009905.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900009905.hg.1 |
ENST00000351266.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009905.hg.1 |
ENST00000351266.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009905.hg.1 |
ENST00000351266.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009905.hg.1 |
ENST00000351266.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009905.hg.1 |
ENST00000351266.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009905.hg.1 |
ENST00000351266.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009905.hg.1 |
ENST00000351266.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009905.hg.1 |
ENST00000351266.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009905.hg.1 |
ENST00000351266.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009905.hg.1 |
ENST00000378980.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009905.hg.1 |
ENST00000378980.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009905.hg.1 |
ENST00000378980.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009905.hg.1 |
ENST00000378980.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009905.hg.1 |
ENST00000378980.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009905.hg.1 |
ENST00000378980.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009905.hg.1 |
ENST00000378980.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009905.hg.1 |
ENST00000378980.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009905.hg.1 |
ENST00000378980.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009905.hg.1 |
ENST00000378980.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009905.hg.1 |
ENST00000378980.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009905.hg.1 |
ENST00000417345.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009905.hg.1 |
ENST00000417345.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009905.hg.1 |
ENST00000417345.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009905.hg.1 |
ENST00000417345.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009905.hg.1 |
ENST00000417345.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009905.hg.1 |
ENST00000417345.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009905.hg.1 |
ENST00000417345.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009905.hg.1 |
ENST00000610543.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009905.hg.1 |
ENST00000610543.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009905.hg.1 |
ENST00000610543.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009905.hg.1 |
ENST00000610543.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009905.hg.1 |
ENST00000610543.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009905.hg.1 |
ENST00000610543.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009905.hg.1 |
ENST00000610543.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009905.hg.1 |
ENST00000610543.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009905.hg.1 |
ENST00000610543.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009905.hg.1 |
ENST00000610543.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009905.hg.1 |
NM_001207011.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009905.hg.1 |
NM_001207011.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009905.hg.1 |
NM_001207011.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009905.hg.1 |
NM_001207011.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009905.hg.1 |
NM_001207011.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009905.hg.1 |
NM_001207011.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009905.hg.1 |
NM_001207011.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009905.hg.1 |
NM_001207011.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009905.hg.1 |
NM_001207011.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009905.hg.1 |
NM_001207011.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009905.hg.1 |
NM_001842.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009905.hg.1 |
NM_001842.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009905.hg.1 |
NM_001842.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009905.hg.1 |
NM_001842.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009905.hg.1 |
NM_001842.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009905.hg.1 |
NM_001842.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009905.hg.1 |
NM_001842.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009905.hg.1 |
NM_001842.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009905.hg.1 |
NM_001842.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009905.hg.1 |
NM_001842.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009905.hg.1 |
NM_147164.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009905.hg.1 |
NM_147164.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009905.hg.1 |
NM_147164.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009905.hg.1 |
NM_147164.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009905.hg.1 |
NM_147164.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009905.hg.1 |
NM_147164.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009905.hg.1 |
NM_147164.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009905.hg.1 |
NM_147164.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009905.hg.1 |
NM_147164.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009905.hg.1 |
NM_147164.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009905.hg.1 |
NM_147164.2 |
Refseq |