Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09022286.hg.1 |
TC09001734.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09022286.hg.1 |
ENST00000277541.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09022286.hg.1 |
NM_017617.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011981.hg.1 |
ENST00000277541.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011981.hg.1 |
ENST00000277541.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011981.hg.1 |
ENST00000277541.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011981.hg.1 |
ENST00000277541.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011981.hg.1 |
ENST00000277541.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011981.hg.1 |
ENST00000277541.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011981.hg.1 |
ENST00000277541.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011981.hg.1 |
ENST00000277541.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011981.hg.1 |
ENST00000277541.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011981.hg.1 |
ENST00000277541.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011981.hg.1 |
ENST00000277541.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011981.hg.1 |
ENST00000277541.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011981.hg.1 |
ENST00000277541.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011981.hg.1 |
ENST00000277541.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011981.hg.1 |
ENST00000277541.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011981.hg.1 |
ENST00000277541.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011981.hg.1 |
ENST00000277541.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011981.hg.1 |
ENST00000277541.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011981.hg.1 |
ENST00000277541.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011981.hg.1 |
ENST00000277541.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011981.hg.1 |
ENST00000277541.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011981.hg.1 |
ENST00000277541.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011981.hg.1 |
ENST00000277541.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011981.hg.1 |
ENST00000277541.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011981.hg.1 |
ENST00000277541.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011981.hg.1 |
ENST00000277541.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011981.hg.1 |
ENST00000277541.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011981.hg.1 |
ENST00000277541.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011981.hg.1 |
ENST00000277541.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011981.hg.1 |
ENST00000277541.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011981.hg.1 |
ENST00000277541.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011981.hg.1 |
ENST00000277541.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011981.hg.1 |
ENST00000277541.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011981.hg.1 |
ENST00000277541.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011981.hg.1 |
ENST00000277541.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011981.hg.1 |
ENST00000277541.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011981.hg.1 |
ENST00000277541.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011981.hg.1 |
ENST00000277541.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011981.hg.1 |
ENST00000277541.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011981.hg.1 |
ENST00000491649.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011981.hg.1 |
ENST00000491649.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011981.hg.1 |
ENST00000491649.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011981.hg.1 |
ENST00000494783.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011981.hg.1 |
ENST00000494783.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011981.hg.1 |
ENST00000494783.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011981.hg.1 |
ENST00000494783.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011981.hg.1 |
NM_017617.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011981.hg.1 |
NM_017617.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011981.hg.1 |
NM_017617.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011981.hg.1 |
NM_017617.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011981.hg.1 |
NM_017617.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011981.hg.1 |
NM_017617.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011981.hg.1 |
NM_017617.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011981.hg.1 |
NM_017617.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011981.hg.1 |
NM_017617.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011981.hg.1 |
NM_017617.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011981.hg.1 |
NM_017617.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011981.hg.1 |
NM_017617.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011981.hg.1 |
NM_017617.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011981.hg.1 |
NM_017617.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011981.hg.1 |
NM_017617.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011981.hg.1 |
NM_017617.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011981.hg.1 |
NM_017617.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011981.hg.1 |
NM_017617.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011981.hg.1 |
NM_017617.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011981.hg.1 |
NM_017617.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011981.hg.1 |
NM_017617.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011981.hg.1 |
NM_017617.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011981.hg.1 |
NM_017617.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011981.hg.1 |
NM_017617.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011981.hg.1 |
NM_017617.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011981.hg.1 |
NM_017617.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011981.hg.1 |
NM_017617.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011981.hg.1 |
NM_017617.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011981.hg.1 |
NM_017617.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011981.hg.1 |
NM_017617.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011981.hg.1 |
NM_017617.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011981.hg.1 |
NM_017617.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011981.hg.1 |
NM_017617.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011981.hg.1 |
NM_017617.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011981.hg.1 |
NM_017617.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011981.hg.1 |
NM_017617.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011981.hg.1 |
NM_017617.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011981.hg.1 |
NM_017617.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900182153.hg.1 |
ENST00000277541.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900182153.hg.1 |
NM_017617.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900182153.hg.1 |
ENST00000277541.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900182153.hg.1 |
NM_017617.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900182153.hg.1 |
TC0900011981.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900011981.hg.1 |
ENST00000277541.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011981.hg.1 |
ENST00000277541.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011981.hg.1 |
ENST00000277541.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011981.hg.1 |
ENST00000277541.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011981.hg.1 |
ENST00000277541.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011981.hg.1 |
ENST00000277541.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011981.hg.1 |
ENST00000277541.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011981.hg.1 |
ENST00000277541.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011981.hg.1 |
ENST00000277541.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011981.hg.1 |
ENST00000277541.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011981.hg.1 |
ENST00000277541.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011981.hg.1 |
ENST00000277541.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011981.hg.1 |
ENST00000277541.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011981.hg.1 |
ENST00000277541.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011981.hg.1 |
ENST00000277541.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011981.hg.1 |
ENST00000277541.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011981.hg.1 |
ENST00000277541.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011981.hg.1 |
ENST00000277541.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011981.hg.1 |
ENST00000277541.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011981.hg.1 |
ENST00000277541.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011981.hg.1 |
ENST00000277541.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011981.hg.1 |
ENST00000277541.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011981.hg.1 |
ENST00000277541.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011981.hg.1 |
ENST00000277541.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011981.hg.1 |
ENST00000277541.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011981.hg.1 |
ENST00000277541.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011981.hg.1 |
ENST00000277541.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011981.hg.1 |
ENST00000277541.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011981.hg.1 |
ENST00000277541.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011981.hg.1 |
ENST00000277541.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011981.hg.1 |
ENST00000277541.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011981.hg.1 |
ENST00000277541.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011981.hg.1 |
ENST00000277541.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011981.hg.1 |
ENST00000277541.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011981.hg.1 |
ENST00000277541.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011981.hg.1 |
ENST00000277541.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011981.hg.1 |
ENST00000277541.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011981.hg.1 |
ENST00000277541.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011981.hg.1 |
ENST00000277541.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011981.hg.1 |
ENST00000491649.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011981.hg.1 |
ENST00000491649.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011981.hg.1 |
ENST00000491649.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011981.hg.1 |
ENST00000494783.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011981.hg.1 |
ENST00000494783.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011981.hg.1 |
ENST00000494783.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011981.hg.1 |
ENST00000494783.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011981.hg.1 |
NM_017617.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011981.hg.1 |
NM_017617.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011981.hg.1 |
NM_017617.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011981.hg.1 |
NM_017617.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011981.hg.1 |
NM_017617.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011981.hg.1 |
NM_017617.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011981.hg.1 |
NM_017617.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011981.hg.1 |
NM_017617.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011981.hg.1 |
NM_017617.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011981.hg.1 |
NM_017617.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011981.hg.1 |
NM_017617.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011981.hg.1 |
NM_017617.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011981.hg.1 |
NM_017617.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011981.hg.1 |
NM_017617.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011981.hg.1 |
NM_017617.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011981.hg.1 |
NM_017617.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011981.hg.1 |
NM_017617.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011981.hg.1 |
NM_017617.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011981.hg.1 |
NM_017617.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011981.hg.1 |
NM_017617.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011981.hg.1 |
NM_017617.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011981.hg.1 |
NM_017617.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011981.hg.1 |
NM_017617.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011981.hg.1 |
NM_017617.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011981.hg.1 |
NM_017617.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011981.hg.1 |
NM_017617.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011981.hg.1 |
NM_017617.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011981.hg.1 |
NM_017617.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011981.hg.1 |
NM_017617.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011981.hg.1 |
NM_017617.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011981.hg.1 |
NM_017617.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011981.hg.1 |
NM_017617.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011981.hg.1 |
NM_017617.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011981.hg.1 |
NM_017617.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011981.hg.1 |
NM_017617.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011981.hg.1 |
NM_017617.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011981.hg.1 |
NM_017617.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011981.hg.1 |
NM_017617.4 |
Refseq |