Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000342435.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000342435.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000342435.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000342435.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000342435.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000342435.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000342435.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000342435.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000342435.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000342435.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000342435.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000342435.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000342435.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000342435.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000342435.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000342435.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000448787.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000448787.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000448787.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000448787.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000448787.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000448787.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000448787.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000448787.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000448787.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000448787.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000462666.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000462666.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000462666.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000462666.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000462666.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000462666.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000462666.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000462666.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000463777.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000463777.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000463777.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000463777.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000463777.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000463777.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000463777.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000468985.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000468985.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000468985.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000468985.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000468985.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000468985.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000468985.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000468985.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000468985.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000468985.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000469266.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000469266.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000469266.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000469266.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000469266.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000469266.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000469266.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000470496.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000470496.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000470496.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000472349.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000472349.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000472349.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000472349.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000472349.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000472349.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000472349.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000472349.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000472349.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000472349.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000472349.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000472349.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000477974.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000477974.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000477974.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000478267.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000478267.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000478267.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000478267.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000478267.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000478267.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000478267.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000478267.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000478267.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000483300.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000483300.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000483300.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000483300.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000483300.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000484560.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000484560.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000484560.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000484560.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000486631.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000486631.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000486631.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000486631.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000486631.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000486631.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000486631.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000487389.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000487389.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000487389.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000487389.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000487389.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000487389.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000487389.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000487389.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000487389.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000487389.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000487389.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000487389.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000487389.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000487389.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000488253.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000488253.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000488253.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000488253.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000488253.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000488253.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000489775.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000489775.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000489775.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000489775.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000489775.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000489775.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000489775.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000489775.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000489775.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000489775.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000490745.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000490745.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000490745.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000490745.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000493432.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000493432.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000493432.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000493432.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000493432.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000493432.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000493432.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000493432.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000493432.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000493432.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000494568.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000494568.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000494568.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000494568.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000494568.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000494568.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000494568.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
ENST00000494568.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
NM_021105.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
NM_021105.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
NM_021105.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
NM_021105.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
NM_021105.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
NM_021105.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
NM_021105.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
NM_021105.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
NM_021105.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
NM_021105.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
NM_021105.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
NM_021105.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
NM_021105.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
NM_021105.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
NM_021105.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012728.hg.1 |
NM_021105.2 |
Refseq |