Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11019437.hg.1 |
ENST00000257832.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11019437.hg.1 |
ENST00000312319.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11019437.hg.1 |
ENST00000528709.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11019437.hg.1 |
NM_001243081.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11019437.hg.1 |
NM_001422.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11019437.hg.1 |
NM_198381.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11019437.hg.1 |
TC11001546.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010514.hg.1 |
ENST00000257832.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010514.hg.1 |
ENST00000257832.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010514.hg.1 |
ENST00000257832.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010514.hg.1 |
ENST00000257832.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010514.hg.1 |
ENST00000257832.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010514.hg.1 |
ENST00000257832.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010514.hg.1 |
ENST00000257832.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010514.hg.1 |
ENST00000257832.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010514.hg.1 |
ENST00000312319.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010514.hg.1 |
ENST00000312319.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010514.hg.1 |
ENST00000312319.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010514.hg.1 |
ENST00000312319.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010514.hg.1 |
ENST00000312319.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010514.hg.1 |
ENST00000312319.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010514.hg.1 |
ENST00000312319.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010514.hg.1 |
ENST00000312319.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010514.hg.1 |
ENST00000312319.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010514.hg.1 |
ENST00000429939.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010514.hg.1 |
ENST00000429939.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010514.hg.1 |
ENST00000429939.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010514.hg.1 |
ENST00000429939.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010514.hg.1 |
ENST00000429939.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010514.hg.1 |
ENST00000528709.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010514.hg.1 |
ENST00000528709.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010514.hg.1 |
ENST00000528709.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010514.hg.1 |
ENST00000528709.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010514.hg.1 |
ENST00000528709.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010514.hg.1 |
ENST00000528709.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010514.hg.1 |
ENST00000532417.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010514.hg.1 |
ENST00000532417.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010514.hg.1 |
ENST00000532417.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010514.hg.1 |
ENST00000532417.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010514.hg.1 |
ENST00000620316.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010514.hg.1 |
ENST00000620316.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010514.hg.1 |
ENST00000620316.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010514.hg.1 |
ENST00000620316.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010514.hg.1 |
ENST00000620316.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010514.hg.1 |
ENST00000620316.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010514.hg.1 |
ENST00000620316.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010514.hg.1 |
ENST00000620316.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010514.hg.1 |
NM_001243080.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010514.hg.1 |
NM_001243080.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010514.hg.1 |
NM_001243080.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010514.hg.1 |
NM_001243080.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010514.hg.1 |
NM_001243080.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010514.hg.1 |
NM_001243080.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010514.hg.1 |
NM_001243081.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010514.hg.1 |
NM_001243081.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010514.hg.1 |
NM_001243081.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010514.hg.1 |
NM_001243081.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010514.hg.1 |
NM_001243081.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010514.hg.1 |
NM_001243081.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010514.hg.1 |
NM_001243081.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010514.hg.1 |
NM_001243081.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010514.hg.1 |
NM_001422.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010514.hg.1 |
NM_001422.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010514.hg.1 |
NM_001422.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010514.hg.1 |
NM_001422.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010514.hg.1 |
NM_001422.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010514.hg.1 |
NM_001422.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010514.hg.1 |
NM_001422.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010514.hg.1 |
NM_001422.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010514.hg.1 |
NM_198381.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010514.hg.1 |
NM_198381.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010514.hg.1 |
NM_198381.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010514.hg.1 |
NM_198381.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010514.hg.1 |
NM_198381.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010514.hg.1 |
NM_198381.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010514.hg.1 |
NM_198381.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010514.hg.1 |
NM_198381.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010514.hg.1 |
NM_198381.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11019441.hg.1 |
ENST00000312319.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11019441.hg.1 |
ENST00000528709.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11019441.hg.1 |
NM_001243081.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11019441.hg.1 |
NM_198381.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11019441.hg.1 |
TC11001546.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1100179753.hg.1 |
ENST00000257832.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100179753.hg.1 |
ENST00000312319.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100179753.hg.1 |
ENST00000528709.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100179753.hg.1 |
ENST00000620316.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100179753.hg.1 |
NM_001243081.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100179753.hg.1 |
NM_001422.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100179753.hg.1 |
NM_198381.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100179753.hg.1 |
ENST00000257832.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100179753.hg.1 |
ENST00000312319.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100179753.hg.1 |
ENST00000528709.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100179753.hg.1 |
ENST00000620316.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100179753.hg.1 |
NM_001243081.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100179753.hg.1 |
NM_001422.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100179753.hg.1 |
NM_198381.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100179753.hg.1 |
TC1100010514.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100010514.hg.1 |
ENST00000257832.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010514.hg.1 |
ENST00000257832.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010514.hg.1 |
ENST00000257832.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010514.hg.1 |
ENST00000257832.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010514.hg.1 |
ENST00000257832.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010514.hg.1 |
ENST00000257832.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010514.hg.1 |
ENST00000257832.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010514.hg.1 |
ENST00000257832.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010514.hg.1 |
ENST00000312319.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010514.hg.1 |
ENST00000312319.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010514.hg.1 |
ENST00000312319.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010514.hg.1 |
ENST00000312319.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010514.hg.1 |
ENST00000312319.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010514.hg.1 |
ENST00000312319.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010514.hg.1 |
ENST00000312319.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010514.hg.1 |
ENST00000312319.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010514.hg.1 |
ENST00000312319.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010514.hg.1 |
ENST00000429939.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010514.hg.1 |
ENST00000429939.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010514.hg.1 |
ENST00000429939.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010514.hg.1 |
ENST00000429939.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010514.hg.1 |
ENST00000429939.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010514.hg.1 |
ENST00000528709.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010514.hg.1 |
ENST00000528709.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010514.hg.1 |
ENST00000528709.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010514.hg.1 |
ENST00000528709.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010514.hg.1 |
ENST00000528709.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010514.hg.1 |
ENST00000528709.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010514.hg.1 |
ENST00000532417.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010514.hg.1 |
ENST00000532417.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010514.hg.1 |
ENST00000532417.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010514.hg.1 |
ENST00000532417.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010514.hg.1 |
ENST00000620316.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010514.hg.1 |
ENST00000620316.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010514.hg.1 |
ENST00000620316.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010514.hg.1 |
ENST00000620316.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010514.hg.1 |
ENST00000620316.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010514.hg.1 |
ENST00000620316.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010514.hg.1 |
ENST00000620316.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010514.hg.1 |
ENST00000620316.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010514.hg.1 |
NM_001243080.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010514.hg.1 |
NM_001243080.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010514.hg.1 |
NM_001243080.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010514.hg.1 |
NM_001243080.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010514.hg.1 |
NM_001243080.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010514.hg.1 |
NM_001243080.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010514.hg.1 |
NM_001243081.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010514.hg.1 |
NM_001243081.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010514.hg.1 |
NM_001243081.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010514.hg.1 |
NM_001243081.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010514.hg.1 |
NM_001243081.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010514.hg.1 |
NM_001243081.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010514.hg.1 |
NM_001243081.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010514.hg.1 |
NM_001243081.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010514.hg.1 |
NM_001422.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010514.hg.1 |
NM_001422.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010514.hg.1 |
NM_001422.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010514.hg.1 |
NM_001422.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010514.hg.1 |
NM_001422.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010514.hg.1 |
NM_001422.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010514.hg.1 |
NM_001422.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010514.hg.1 |
NM_001422.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010514.hg.1 |
NM_198381.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010514.hg.1 |
NM_198381.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010514.hg.1 |
NM_198381.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010514.hg.1 |
NM_198381.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010514.hg.1 |
NM_198381.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010514.hg.1 |
NM_198381.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010514.hg.1 |
NM_198381.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010514.hg.1 |
NM_198381.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010514.hg.1 |
NM_198381.1 |
Refseq |