Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010636.hg.1 |
ENST00000290953.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010636.hg.1 |
ENST00000290953.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010636.hg.1 |
ENST00000290953.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010636.hg.1 |
ENST00000290953.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010636.hg.1 |
ENST00000290953.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010636.hg.1 |
NM_001138.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010636.hg.1 |
NM_001138.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010636.hg.1 |
NM_001138.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010636.hg.1 |
NM_001138.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600010636.hg.1 |
NM_001138.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16016567.hg.1 |
ENST00000290953.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16016567.hg.1 |
NM_001138.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16016567.hg.1 |
TC16001191.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR16016568.hg.1 |
ENST00000290953.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16016568.hg.1 |
NM_001138.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16016568.hg.1 |
TC16001191.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1600182007.hg.1 |
ENST00000290953.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600182007.hg.1 |
NM_001138.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600182007.hg.1 |
ENST00000290953.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600182007.hg.1 |
NM_001138.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600182007.hg.1 |
TC1600010636.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1600010636.hg.1 |
ENST00000290953.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010636.hg.1 |
ENST00000290953.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010636.hg.1 |
ENST00000290953.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010636.hg.1 |
ENST00000290953.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010636.hg.1 |
ENST00000290953.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600010636.hg.1 |
NM_001138.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010636.hg.1 |
NM_001138.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010636.hg.1 |
NM_001138.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010636.hg.1 |
NM_001138.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600010636.hg.1 |
NM_001138.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600182009.hg.1 |
ENST00000290953.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600182009.hg.1 |
NM_001138.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600182009.hg.1 |
ENST00000290953.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600182009.hg.1 |
NM_001138.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600182009.hg.1 |
TC1600010636.hg.1 |
Clariom D TC |