Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR16002570.hg.1 |
ENST00000311895.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16002570.hg.1 |
ENST00000389138.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16002570.hg.1 |
ENST00000439007.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16002570.hg.1 |
NM_005236.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16002570.hg.1 |
TC16000170.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR16002569.hg.1 |
ENST00000311895.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16002569.hg.1 |
ENST00000389138.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16002569.hg.1 |
ENST00000439007.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16002569.hg.1 |
NM_005236.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16002569.hg.1 |
TC16000170.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006976.hg.1 |
ENST00000311895.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006976.hg.1 |
ENST00000311895.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006976.hg.1 |
ENST00000311895.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006976.hg.1 |
ENST00000311895.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006976.hg.1 |
ENST00000311895.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006976.hg.1 |
ENST00000311895.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006976.hg.1 |
ENST00000311895.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006976.hg.1 |
ENST00000311895.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006976.hg.1 |
ENST00000311895.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006976.hg.1 |
ENST00000311895.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006976.hg.1 |
ENST00000311895.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006976.hg.1 |
ENST00000311895.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006976.hg.1 |
ENST00000311895.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006976.hg.1 |
ENST00000311895.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006976.hg.1 |
ENST00000311895.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006976.hg.1 |
ENST00000311895.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006976.hg.1 |
ENST00000311895.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006976.hg.1 |
ENST00000311895.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006976.hg.1 |
ENST00000389138.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006976.hg.1 |
ENST00000389138.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006976.hg.1 |
ENST00000389138.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006976.hg.1 |
ENST00000389138.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006976.hg.1 |
ENST00000389138.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006976.hg.1 |
ENST00000389138.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006976.hg.1 |
ENST00000389138.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006976.hg.1 |
ENST00000389138.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006976.hg.1 |
ENST00000389138.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006976.hg.1 |
ENST00000389138.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006976.hg.1 |
ENST00000389138.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006976.hg.1 |
ENST00000389138.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006976.hg.1 |
ENST00000462862.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006976.hg.1 |
ENST00000462862.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006976.hg.1 |
ENST00000462862.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006976.hg.1 |
ENST00000462862.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006976.hg.1 |
ENST00000573018.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006976.hg.1 |
ENST00000573018.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006976.hg.1 |
ENST00000573018.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006976.hg.1 |
ENST00000574194.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006976.hg.1 |
ENST00000574194.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006976.hg.1 |
ENST00000574194.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006976.hg.1 |
ENST00000574194.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006976.hg.1 |
ENST00000574781.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006976.hg.1 |
ENST00000574781.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006976.hg.1 |
ENST00000574781.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006976.hg.1 |
ENST00000574781.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006976.hg.1 |
ENST00000574781.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006976.hg.1 |
ENST00000575156.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006976.hg.1 |
ENST00000575156.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006976.hg.1 |
ENST00000575156.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006976.hg.1 |
ENST00000575156.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006976.hg.1 |
ENST00000575156.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006976.hg.1 |
ENST00000575156.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006976.hg.1 |
ENST00000575156.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006976.hg.1 |
ENST00000575156.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006976.hg.1 |
ENST00000575156.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006976.hg.1 |
ENST00000575156.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006976.hg.1 |
ENST00000575156.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006976.hg.1 |
ENST00000576348.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006976.hg.1 |
ENST00000576348.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006976.hg.1 |
ENST00000576348.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006976.hg.1 |
ENST00000576348.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006976.hg.1 |
NM_005236.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006976.hg.1 |
NM_005236.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006976.hg.1 |
NM_005236.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006976.hg.1 |
NM_005236.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006976.hg.1 |
NM_005236.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006976.hg.1 |
NM_005236.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006976.hg.1 |
NM_005236.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006976.hg.1 |
NM_005236.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006976.hg.1 |
NM_005236.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006976.hg.1 |
NM_005236.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006976.hg.1 |
NM_005236.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006976.hg.1 |
NM_005236.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006976.hg.1 |
NM_005236.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006976.hg.1 |
NM_005236.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006976.hg.1 |
NM_005236.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006976.hg.1 |
NM_005236.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006976.hg.1 |
NM_005236.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006976.hg.1 |
NM_005236.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600149394.hg.1 |
ENST00000311895.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600149394.hg.1 |
ENST00000575156.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600149394.hg.1 |
ENST00000576348.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600149394.hg.1 |
NM_005236.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600149394.hg.1 |
ENST00000311895.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600149394.hg.1 |
ENST00000575156.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600149394.hg.1 |
ENST00000576348.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600149394.hg.1 |
NM_005236.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600149394.hg.1 |
TC1600006976.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1600006976.hg.1 |
ENST00000311895.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006976.hg.1 |
ENST00000311895.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006976.hg.1 |
ENST00000311895.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006976.hg.1 |
ENST00000311895.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006976.hg.1 |
ENST00000311895.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006976.hg.1 |
ENST00000311895.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006976.hg.1 |
ENST00000311895.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006976.hg.1 |
ENST00000311895.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006976.hg.1 |
ENST00000311895.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006976.hg.1 |
ENST00000311895.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006976.hg.1 |
ENST00000311895.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006976.hg.1 |
ENST00000311895.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006976.hg.1 |
ENST00000311895.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006976.hg.1 |
ENST00000311895.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006976.hg.1 |
ENST00000311895.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006976.hg.1 |
ENST00000311895.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006976.hg.1 |
ENST00000311895.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006976.hg.1 |
ENST00000311895.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006976.hg.1 |
ENST00000389138.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006976.hg.1 |
ENST00000389138.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006976.hg.1 |
ENST00000389138.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006976.hg.1 |
ENST00000389138.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006976.hg.1 |
ENST00000389138.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006976.hg.1 |
ENST00000389138.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006976.hg.1 |
ENST00000389138.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006976.hg.1 |
ENST00000389138.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006976.hg.1 |
ENST00000389138.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006976.hg.1 |
ENST00000389138.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006976.hg.1 |
ENST00000389138.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006976.hg.1 |
ENST00000389138.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006976.hg.1 |
ENST00000462862.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006976.hg.1 |
ENST00000462862.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006976.hg.1 |
ENST00000462862.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006976.hg.1 |
ENST00000462862.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006976.hg.1 |
ENST00000573018.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006976.hg.1 |
ENST00000573018.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006976.hg.1 |
ENST00000573018.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006976.hg.1 |
ENST00000574194.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006976.hg.1 |
ENST00000574194.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006976.hg.1 |
ENST00000574194.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006976.hg.1 |
ENST00000574194.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006976.hg.1 |
ENST00000574781.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006976.hg.1 |
ENST00000574781.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006976.hg.1 |
ENST00000574781.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006976.hg.1 |
ENST00000574781.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006976.hg.1 |
ENST00000574781.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006976.hg.1 |
ENST00000575156.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006976.hg.1 |
ENST00000575156.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006976.hg.1 |
ENST00000575156.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006976.hg.1 |
ENST00000575156.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006976.hg.1 |
ENST00000575156.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006976.hg.1 |
ENST00000575156.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006976.hg.1 |
ENST00000575156.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006976.hg.1 |
ENST00000575156.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006976.hg.1 |
ENST00000575156.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006976.hg.1 |
ENST00000575156.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006976.hg.1 |
ENST00000575156.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006976.hg.1 |
ENST00000576348.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006976.hg.1 |
ENST00000576348.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006976.hg.1 |
ENST00000576348.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006976.hg.1 |
ENST00000576348.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006976.hg.1 |
NM_005236.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006976.hg.1 |
NM_005236.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006976.hg.1 |
NM_005236.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006976.hg.1 |
NM_005236.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006976.hg.1 |
NM_005236.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006976.hg.1 |
NM_005236.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006976.hg.1 |
NM_005236.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006976.hg.1 |
NM_005236.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006976.hg.1 |
NM_005236.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006976.hg.1 |
NM_005236.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006976.hg.1 |
NM_005236.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006976.hg.1 |
NM_005236.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006976.hg.1 |
NM_005236.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006976.hg.1 |
NM_005236.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006976.hg.1 |
NM_005236.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006976.hg.1 |
NM_005236.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006976.hg.1 |
NM_005236.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006976.hg.1 |
NM_005236.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600149393.hg.1 |
ENST00000311895.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600149393.hg.1 |
ENST00000575156.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600149393.hg.1 |
NM_005236.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600149393.hg.1 |
ENST00000311895.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600149393.hg.1 |
ENST00000575156.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600149393.hg.1 |
NM_005236.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600149393.hg.1 |
TC1600006976.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600149395.hg.1 |
ENST00000311895.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600149395.hg.1 |
ENST00000575156.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600149395.hg.1 |
ENST00000576348.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600149395.hg.1 |
NM_005236.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600149395.hg.1 |
ENST00000311895.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600149395.hg.1 |
ENST00000575156.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600149395.hg.1 |
ENST00000576348.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600149395.hg.1 |
NM_005236.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600149395.hg.1 |
TC1600006976.hg.1 |
Clariom D TC |