Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012313.hg.1 |
ENST00000333676.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012313.hg.1 |
ENST00000333676.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012313.hg.1 |
ENST00000333676.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012313.hg.1 |
ENST00000333676.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012313.hg.1 |
ENST00000333676.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012313.hg.1 |
ENST00000333676.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012313.hg.1 |
ENST00000333676.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012313.hg.1 |
ENST00000541223.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012313.hg.1 |
ENST00000541223.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012313.hg.1 |
ENST00000541223.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012313.hg.1 |
ENST00000541223.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012313.hg.1 |
ENST00000541223.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012313.hg.1 |
ENST00000541223.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012313.hg.1 |
NM_002949.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012313.hg.1 |
NM_002949.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012313.hg.1 |
NM_002949.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012313.hg.1 |
NM_002949.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012313.hg.1 |
NM_002949.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012313.hg.1 |
NM_002949.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012313.hg.1 |
NM_002949.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17012316.hg.1 |
TC17002882.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR17012316.hg.1 |
ENST00000332396.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17012316.hg.1 |
ENST00000333676.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17012316.hg.1 |
NM_002949.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700204030.hg.1 |
ENST00000333676.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700204030.hg.1 |
ENST00000541223.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700204030.hg.1 |
NM_002949.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700204030.hg.1 |
ENST00000333676.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700204030.hg.1 |
ENST00000541223.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700204030.hg.1 |
NM_002949.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700204030.hg.1 |
TC1700012313.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1700012313.hg.1 |
ENST00000333676.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012313.hg.1 |
ENST00000333676.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012313.hg.1 |
ENST00000333676.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012313.hg.1 |
ENST00000333676.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012313.hg.1 |
ENST00000333676.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012313.hg.1 |
ENST00000333676.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012313.hg.1 |
ENST00000333676.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012313.hg.1 |
ENST00000541223.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012313.hg.1 |
ENST00000541223.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012313.hg.1 |
ENST00000541223.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012313.hg.1 |
ENST00000541223.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012313.hg.1 |
ENST00000541223.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012313.hg.1 |
ENST00000541223.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012313.hg.1 |
NM_002949.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012313.hg.1 |
NM_002949.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012313.hg.1 |
NM_002949.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012313.hg.1 |
NM_002949.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012313.hg.1 |
NM_002949.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012313.hg.1 |
NM_002949.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012313.hg.1 |
NM_002949.3 |
Refseq |