Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007713.hg.1 |
TC02000502.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007713.hg.1 |
ENST00000282111.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02007713.hg.1 |
NM_031283.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008236.hg.1 |
ENST00000282111.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008236.hg.1 |
ENST00000282111.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008236.hg.1 |
ENST00000282111.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008236.hg.1 |
ENST00000282111.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008236.hg.1 |
ENST00000282111.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008236.hg.1 |
ENST00000282111.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008236.hg.1 |
ENST00000282111.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008236.hg.1 |
ENST00000282111.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008236.hg.1 |
ENST00000282111.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008236.hg.1 |
ENST00000282111.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008236.hg.1 |
ENST00000282111.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008236.hg.1 |
ENST00000282111.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008236.hg.1 |
ENST00000282111.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008236.hg.1 |
ENST00000282111.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008236.hg.1 |
ENST00000282111.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008236.hg.1 |
ENST00000282111.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008236.hg.1 |
ENST00000282111.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008236.hg.1 |
ENST00000442813.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008236.hg.1 |
ENST00000442813.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008236.hg.1 |
ENST00000442813.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008236.hg.1 |
ENST00000442813.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008236.hg.1 |
ENST00000442813.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008236.hg.1 |
ENST00000490744.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008236.hg.1 |
ENST00000490744.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008236.hg.1 |
ENST00000490744.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008236.hg.1 |
ENST00000490744.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008236.hg.1 |
ENST00000494519.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008236.hg.1 |
ENST00000494519.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008236.hg.1 |
NM_031283.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008236.hg.1 |
NM_031283.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008236.hg.1 |
NM_031283.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008236.hg.1 |
NM_031283.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008236.hg.1 |
NM_031283.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008236.hg.1 |
NM_031283.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008236.hg.1 |
NM_031283.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008236.hg.1 |
NM_031283.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008236.hg.1 |
NM_031283.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008236.hg.1 |
NM_031283.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008236.hg.1 |
NM_031283.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008236.hg.1 |
NM_031283.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008236.hg.1 |
NM_031283.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008236.hg.1 |
NM_031283.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008236.hg.1 |
NM_031283.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008236.hg.1 |
NM_031283.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200157022.hg.1 |
ENST00000282111.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200157022.hg.1 |
NM_031283.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200157022.hg.1 |
ENST00000282111.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200157022.hg.1 |
NM_031283.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200157022.hg.1 |
TC0200008236.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200008236.hg.1 |
ENST00000282111.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008236.hg.1 |
ENST00000282111.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008236.hg.1 |
ENST00000282111.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008236.hg.1 |
ENST00000282111.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008236.hg.1 |
ENST00000282111.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008236.hg.1 |
ENST00000282111.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008236.hg.1 |
ENST00000282111.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008236.hg.1 |
ENST00000282111.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008236.hg.1 |
ENST00000282111.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008236.hg.1 |
ENST00000282111.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008236.hg.1 |
ENST00000282111.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008236.hg.1 |
ENST00000282111.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008236.hg.1 |
ENST00000282111.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008236.hg.1 |
ENST00000282111.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008236.hg.1 |
ENST00000282111.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008236.hg.1 |
ENST00000282111.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008236.hg.1 |
ENST00000282111.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008236.hg.1 |
ENST00000442813.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008236.hg.1 |
ENST00000442813.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008236.hg.1 |
ENST00000442813.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008236.hg.1 |
ENST00000442813.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008236.hg.1 |
ENST00000442813.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008236.hg.1 |
ENST00000490744.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008236.hg.1 |
ENST00000490744.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008236.hg.1 |
ENST00000490744.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008236.hg.1 |
ENST00000490744.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008236.hg.1 |
ENST00000494519.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008236.hg.1 |
ENST00000494519.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008236.hg.1 |
NM_031283.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008236.hg.1 |
NM_031283.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008236.hg.1 |
NM_031283.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008236.hg.1 |
NM_031283.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008236.hg.1 |
NM_031283.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008236.hg.1 |
NM_031283.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008236.hg.1 |
NM_031283.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008236.hg.1 |
NM_031283.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008236.hg.1 |
NM_031283.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008236.hg.1 |
NM_031283.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008236.hg.1 |
NM_031283.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008236.hg.1 |
NM_031283.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008236.hg.1 |
NM_031283.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008236.hg.1 |
NM_031283.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008236.hg.1 |
NM_031283.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008236.hg.1 |
NM_031283.2 |
Refseq |