Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17021051.hg.1 |
ENST00000360085.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17021051.hg.1 |
NM_004160.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17021051.hg.1 |
TC17001568.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010807.hg.1 |
ENST00000360085.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010807.hg.1 |
ENST00000360085.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010807.hg.1 |
ENST00000360085.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010807.hg.1 |
ENST00000360085.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010807.hg.1 |
ENST00000360085.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010807.hg.1 |
ENST00000360085.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010807.hg.1 |
ENST00000360085.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010807.hg.1 |
ENST00000360085.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010807.hg.1 |
ENST00000360085.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010807.hg.1 |
ENST00000360085.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010807.hg.1 |
ENST00000592796.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010807.hg.1 |
ENST00000592796.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010807.hg.1 |
ENST00000592796.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010807.hg.1 |
ENST00000592796.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010807.hg.1 |
ENST00000592796.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010807.hg.1 |
ENST00000592796.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010807.hg.1 |
NM_004160.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010807.hg.1 |
NM_004160.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010807.hg.1 |
NM_004160.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010807.hg.1 |
NM_004160.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010807.hg.1 |
NM_004160.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010807.hg.1 |
NM_004160.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010807.hg.1 |
NM_004160.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010807.hg.1 |
NM_004160.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010807.hg.1 |
NM_004160.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010807.hg.1 |
NM_004160.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700186281.hg.1 |
ENST00000360085.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700186281.hg.1 |
ENST00000592796.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700186281.hg.1 |
NM_004160.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700186281.hg.1 |
ENST00000360085.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700186281.hg.1 |
ENST00000592796.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700186281.hg.1 |
NM_004160.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700186281.hg.1 |
TC1700010807.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1700010807.hg.1 |
ENST00000360085.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010807.hg.1 |
ENST00000360085.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010807.hg.1 |
ENST00000360085.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010807.hg.1 |
ENST00000360085.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010807.hg.1 |
ENST00000360085.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010807.hg.1 |
ENST00000360085.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010807.hg.1 |
ENST00000360085.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010807.hg.1 |
ENST00000360085.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010807.hg.1 |
ENST00000360085.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010807.hg.1 |
ENST00000360085.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010807.hg.1 |
ENST00000592796.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010807.hg.1 |
ENST00000592796.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010807.hg.1 |
ENST00000592796.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010807.hg.1 |
ENST00000592796.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010807.hg.1 |
ENST00000592796.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010807.hg.1 |
ENST00000592796.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010807.hg.1 |
NM_004160.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010807.hg.1 |
NM_004160.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010807.hg.1 |
NM_004160.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010807.hg.1 |
NM_004160.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010807.hg.1 |
NM_004160.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010807.hg.1 |
NM_004160.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010807.hg.1 |
NM_004160.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010807.hg.1 |
NM_004160.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010807.hg.1 |
NM_004160.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010807.hg.1 |
NM_004160.5 |
Refseq |