Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR15013019.hg.1 |
ENST00000220429.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15013019.hg.1 |
ENST00000359031.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15013019.hg.1 |
ENST00000380877.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15013019.hg.1 |
ENST00000396464.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15013019.hg.1 |
ENST00000429662.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15013019.hg.1 |
ENST00000543881.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15013019.hg.1 |
ENST00000560825.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15013019.hg.1 |
NM_002041.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15013019.hg.1 |
NM_005254.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15013019.hg.1 |
NM_016654.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15013019.hg.1 |
NM_016655.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15013019.hg.1 |
NM_181427.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15013019.hg.1 |
TC15001350.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1500168953.hg.1 |
ENST00000220429.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500168953.hg.1 |
ENST00000359031.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500168953.hg.1 |
ENST00000380877.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500168953.hg.1 |
ENST00000396464.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500168953.hg.1 |
ENST00000429662.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500168953.hg.1 |
ENST00000543881.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500168953.hg.1 |
ENST00000560825.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500168953.hg.1 |
ENST00000561010.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500168953.hg.1 |
NM_002041.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500168953.hg.1 |
NM_005254.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500168953.hg.1 |
NM_016654.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500168953.hg.1 |
NM_016655.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500168953.hg.1 |
NM_181427.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500168953.hg.1 |
ENST00000220429.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500168953.hg.1 |
ENST00000359031.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500168953.hg.1 |
ENST00000380877.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500168953.hg.1 |
ENST00000396464.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500168953.hg.1 |
ENST00000429662.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500168953.hg.1 |
ENST00000543881.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500168953.hg.1 |
ENST00000560825.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500168953.hg.1 |
ENST00000561010.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500168953.hg.1 |
NM_002041.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500168953.hg.1 |
NM_005254.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500168953.hg.1 |
NM_016654.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500168953.hg.1 |
NM_016655.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500168953.hg.1 |
NM_181427.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500168953.hg.1 |
TC1500009386.hg.1 |
Clariom D TC |