Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR15012230.hg.1 |
TC15002773.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR15012230.hg.1 |
ENST00000319359.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15012230.hg.1 |
NM_025165.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010862.hg.1 |
ENST00000319359.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010862.hg.1 |
ENST00000319359.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010862.hg.1 |
ENST00000319359.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010862.hg.1 |
ENST00000319359.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010862.hg.1 |
ENST00000319359.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010862.hg.1 |
ENST00000319359.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010862.hg.1 |
ENST00000319359.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010862.hg.1 |
ENST00000319359.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010862.hg.1 |
ENST00000319359.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010862.hg.1 |
ENST00000319359.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010862.hg.1 |
ENST00000319359.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010862.hg.1 |
ENST00000319359.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010862.hg.1 |
ENST00000319359.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010862.hg.1 |
ENST00000319359.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010862.hg.1 |
ENST00000319359.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010862.hg.1 |
ENST00000319359.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010862.hg.1 |
ENST00000319359.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010862.hg.1 |
ENST00000319359.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010862.hg.1 |
ENST00000319359.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010862.hg.1 |
ENST00000433927.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010862.hg.1 |
ENST00000433927.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010862.hg.1 |
ENST00000433927.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010862.hg.1 |
ENST00000433927.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010862.hg.1 |
ENST00000433927.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010862.hg.1 |
ENST00000433927.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010862.hg.1 |
ENST00000433927.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010862.hg.1 |
ENST00000467869.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010862.hg.1 |
ENST00000467869.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010862.hg.1 |
ENST00000467869.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010862.hg.1 |
ENST00000467869.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010862.hg.1 |
ENST00000467869.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010862.hg.1 |
ENST00000467869.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010862.hg.1 |
ENST00000467869.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010862.hg.1 |
ENST00000467869.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010862.hg.1 |
ENST00000467869.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010862.hg.1 |
ENST00000467869.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010862.hg.1 |
ENST00000467869.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010862.hg.1 |
ENST00000467869.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010862.hg.1 |
ENST00000467869.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010862.hg.1 |
ENST00000467869.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010862.hg.1 |
ENST00000476335.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010862.hg.1 |
ENST00000476335.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010862.hg.1 |
ENST00000476335.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010862.hg.1 |
ENST00000476335.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010862.hg.1 |
ENST00000476335.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010862.hg.1 |
ENST00000476335.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010862.hg.1 |
ENST00000486851.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010862.hg.1 |
ENST00000486851.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010862.hg.1 |
ENST00000486851.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010862.hg.1 |
ENST00000486851.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010862.hg.1 |
ENST00000486851.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010862.hg.1 |
ENST00000486851.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010862.hg.1 |
ENST00000486851.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010862.hg.1 |
ENST00000497465.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010862.hg.1 |
ENST00000497465.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010862.hg.1 |
ENST00000497465.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010862.hg.1 |
ENST00000497465.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010862.hg.1 |
ENST00000497465.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010862.hg.1 |
ENST00000497465.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010862.hg.1 |
ENST00000497465.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010862.hg.1 |
ENST00000497465.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010862.hg.1 |
ENST00000497465.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010862.hg.1 |
ENST00000497530.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010862.hg.1 |
ENST00000497530.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010862.hg.1 |
ENST00000497530.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010862.hg.1 |
ENST00000497530.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010862.hg.1 |
ENST00000497530.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010862.hg.1 |
ENST00000497530.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010862.hg.1 |
ENST00000497530.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010862.hg.1 |
ENST00000497700.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010862.hg.1 |
ENST00000497700.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010862.hg.1 |
ENST00000497700.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010862.hg.1 |
ENST00000497700.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010862.hg.1 |
ENST00000497700.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010862.hg.1 |
ENST00000497700.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010862.hg.1 |
NM_025165.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010862.hg.1 |
NM_025165.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010862.hg.1 |
NM_025165.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010862.hg.1 |
NM_025165.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010862.hg.1 |
NM_025165.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010862.hg.1 |
NM_025165.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010862.hg.1 |
NM_025165.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010862.hg.1 |
NM_025165.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010862.hg.1 |
NM_025165.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010862.hg.1 |
NM_025165.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010862.hg.1 |
NM_025165.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010862.hg.1 |
NM_025165.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010862.hg.1 |
NM_025165.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010862.hg.1 |
NM_025165.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010862.hg.1 |
NM_025165.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010862.hg.1 |
NM_025165.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010862.hg.1 |
NM_025165.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010862.hg.1 |
NM_025165.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500184492.hg.1 |
ENST00000319359.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500184492.hg.1 |
ENST00000433927.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500184492.hg.1 |
ENST00000467869.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500184492.hg.1 |
ENST00000476335.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500184492.hg.1 |
ENST00000486851.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500184492.hg.1 |
ENST00000497530.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500184492.hg.1 |
NM_025165.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500184492.hg.1 |
ENST00000319359.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500184492.hg.1 |
ENST00000433927.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500184492.hg.1 |
ENST00000467869.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500184492.hg.1 |
ENST00000476335.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500184492.hg.1 |
ENST00000486851.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500184492.hg.1 |
ENST00000497530.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500184492.hg.1 |
NM_025165.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500184492.hg.1 |
TC1500010862.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1500010862.hg.1 |
ENST00000319359.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010862.hg.1 |
ENST00000319359.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010862.hg.1 |
ENST00000319359.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010862.hg.1 |
ENST00000319359.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010862.hg.1 |
ENST00000319359.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010862.hg.1 |
ENST00000319359.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010862.hg.1 |
ENST00000319359.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010862.hg.1 |
ENST00000319359.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010862.hg.1 |
ENST00000319359.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010862.hg.1 |
ENST00000319359.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010862.hg.1 |
ENST00000319359.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010862.hg.1 |
ENST00000319359.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010862.hg.1 |
ENST00000319359.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010862.hg.1 |
ENST00000319359.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010862.hg.1 |
ENST00000319359.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010862.hg.1 |
ENST00000319359.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010862.hg.1 |
ENST00000319359.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010862.hg.1 |
ENST00000319359.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010862.hg.1 |
ENST00000319359.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010862.hg.1 |
ENST00000433927.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010862.hg.1 |
ENST00000433927.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010862.hg.1 |
ENST00000433927.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010862.hg.1 |
ENST00000433927.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010862.hg.1 |
ENST00000433927.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010862.hg.1 |
ENST00000433927.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010862.hg.1 |
ENST00000433927.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010862.hg.1 |
ENST00000467869.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010862.hg.1 |
ENST00000467869.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010862.hg.1 |
ENST00000467869.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010862.hg.1 |
ENST00000467869.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010862.hg.1 |
ENST00000467869.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010862.hg.1 |
ENST00000467869.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010862.hg.1 |
ENST00000467869.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010862.hg.1 |
ENST00000467869.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010862.hg.1 |
ENST00000467869.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010862.hg.1 |
ENST00000467869.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010862.hg.1 |
ENST00000467869.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010862.hg.1 |
ENST00000467869.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010862.hg.1 |
ENST00000467869.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010862.hg.1 |
ENST00000467869.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010862.hg.1 |
ENST00000476335.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010862.hg.1 |
ENST00000476335.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010862.hg.1 |
ENST00000476335.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010862.hg.1 |
ENST00000476335.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010862.hg.1 |
ENST00000476335.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010862.hg.1 |
ENST00000476335.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010862.hg.1 |
ENST00000486851.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010862.hg.1 |
ENST00000486851.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010862.hg.1 |
ENST00000486851.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010862.hg.1 |
ENST00000486851.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010862.hg.1 |
ENST00000486851.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010862.hg.1 |
ENST00000486851.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010862.hg.1 |
ENST00000486851.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010862.hg.1 |
ENST00000497465.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010862.hg.1 |
ENST00000497465.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010862.hg.1 |
ENST00000497465.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010862.hg.1 |
ENST00000497465.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010862.hg.1 |
ENST00000497465.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010862.hg.1 |
ENST00000497465.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010862.hg.1 |
ENST00000497465.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010862.hg.1 |
ENST00000497465.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010862.hg.1 |
ENST00000497465.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010862.hg.1 |
ENST00000497530.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010862.hg.1 |
ENST00000497530.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010862.hg.1 |
ENST00000497530.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010862.hg.1 |
ENST00000497530.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010862.hg.1 |
ENST00000497530.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010862.hg.1 |
ENST00000497530.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010862.hg.1 |
ENST00000497530.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010862.hg.1 |
ENST00000497700.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010862.hg.1 |
ENST00000497700.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010862.hg.1 |
ENST00000497700.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010862.hg.1 |
ENST00000497700.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010862.hg.1 |
ENST00000497700.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010862.hg.1 |
ENST00000497700.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010862.hg.1 |
NM_025165.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010862.hg.1 |
NM_025165.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010862.hg.1 |
NM_025165.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010862.hg.1 |
NM_025165.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010862.hg.1 |
NM_025165.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010862.hg.1 |
NM_025165.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010862.hg.1 |
NM_025165.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010862.hg.1 |
NM_025165.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010862.hg.1 |
NM_025165.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010862.hg.1 |
NM_025165.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010862.hg.1 |
NM_025165.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010862.hg.1 |
NM_025165.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010862.hg.1 |
NM_025165.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010862.hg.1 |
NM_025165.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010862.hg.1 |
NM_025165.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010862.hg.1 |
NM_025165.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010862.hg.1 |
NM_025165.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010862.hg.1 |
NM_025165.2 |
Refseq |