Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02014010.hg.1 |
TC02000960.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02014010.hg.1 |
ENST00000389757.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02014010.hg.1 |
ENST00000389759.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02014010.hg.1 |
ENST00000428353.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02014010.hg.1 |
NM_001005476.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02014010.hg.1 |
NM_003628.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02014014.hg.1 |
TC02000960.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02014014.hg.1 |
ENST00000389756.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02014014.hg.1 |
ENST00000389757.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02014014.hg.1 |
ENST00000389759.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02014014.hg.1 |
ENST00000428353.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02014014.hg.1 |
NM_001005476.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02014014.hg.1 |
NM_003628.4 |
Refseq |