Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015061.hg.1 |
ENST00000370131.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015061.hg.1 |
ENST00000370131.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015061.hg.1 |
ENST00000370131.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015061.hg.1 |
ENST00000370131.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015061.hg.1 |
ENST00000370131.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015061.hg.1 |
ENST00000370131.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015061.hg.1 |
ENST00000370131.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015061.hg.1 |
ENST00000370131.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015061.hg.1 |
ENST00000370131.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015061.hg.1 |
ENST00000370131.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015061.hg.1 |
ENST00000370132.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015061.hg.1 |
ENST00000370132.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015061.hg.1 |
ENST00000370132.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015061.hg.1 |
ENST00000370132.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015061.hg.1 |
ENST00000370132.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015061.hg.1 |
ENST00000370132.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015061.hg.1 |
ENST00000370132.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015061.hg.1 |
ENST00000370132.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015061.hg.1 |
ENST00000370132.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015061.hg.1 |
ENST00000370132.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015061.hg.1 |
ENST00000370132.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015061.hg.1 |
ENST00000370132.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015061.hg.1 |
ENST00000370132.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015061.hg.1 |
NM_001918.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015061.hg.1 |
NM_001918.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015061.hg.1 |
NM_001918.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015061.hg.1 |
NM_001918.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015061.hg.1 |
NM_001918.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015061.hg.1 |
NM_001918.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015061.hg.1 |
NM_001918.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015061.hg.1 |
NM_001918.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015061.hg.1 |
NM_001918.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015061.hg.1 |
NM_001918.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015061.hg.1 |
NM_001918.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015061.hg.1 |
NM_001918.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015061.hg.1 |
NM_001918.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012031.hg.1 |
ENST00000496068.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01045361.hg.1 |
TC01002918.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01045361.hg.1 |
ENST00000370131.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01045361.hg.1 |
ENST00000370132.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01045361.hg.1 |
ENST00000543138.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01045361.hg.1 |
NM_001918.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01045363.hg.1 |
TC01002918.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01045363.hg.1 |
ENST00000370131.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01045363.hg.1 |
ENST00000370132.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01045363.hg.1 |
ENST00000543138.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01045363.hg.1 |
NM_001918.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100212217.hg.1 |
ENST00000370131.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100212217.hg.1 |
ENST00000370132.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100212217.hg.1 |
NM_001918.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100212217.hg.1 |
ENST00000370131.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100212217.hg.1 |
ENST00000370132.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100212217.hg.1 |
NM_001918.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100212217.hg.1 |
TC0100015061.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100015061.hg.1 |
ENST00000370131.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015061.hg.1 |
ENST00000370131.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015061.hg.1 |
ENST00000370131.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015061.hg.1 |
ENST00000370131.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015061.hg.1 |
ENST00000370131.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015061.hg.1 |
ENST00000370131.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015061.hg.1 |
ENST00000370131.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015061.hg.1 |
ENST00000370131.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015061.hg.1 |
ENST00000370131.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015061.hg.1 |
ENST00000370131.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015061.hg.1 |
ENST00000370132.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015061.hg.1 |
ENST00000370132.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015061.hg.1 |
ENST00000370132.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015061.hg.1 |
ENST00000370132.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015061.hg.1 |
ENST00000370132.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015061.hg.1 |
ENST00000370132.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015061.hg.1 |
ENST00000370132.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015061.hg.1 |
ENST00000370132.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015061.hg.1 |
ENST00000370132.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015061.hg.1 |
ENST00000370132.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015061.hg.1 |
ENST00000370132.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015061.hg.1 |
ENST00000370132.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015061.hg.1 |
ENST00000370132.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015061.hg.1 |
NM_001918.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015061.hg.1 |
NM_001918.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015061.hg.1 |
NM_001918.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015061.hg.1 |
NM_001918.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015061.hg.1 |
NM_001918.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015061.hg.1 |
NM_001918.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015061.hg.1 |
NM_001918.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015061.hg.1 |
NM_001918.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015061.hg.1 |
NM_001918.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015061.hg.1 |
NM_001918.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015061.hg.1 |
NM_001918.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015061.hg.1 |
NM_001918.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015061.hg.1 |
NM_001918.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100212216.hg.1 |
ENST00000370131.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100212216.hg.1 |
ENST00000370132.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100212216.hg.1 |
NM_001918.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100212216.hg.1 |
ENST00000370131.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100212216.hg.1 |
ENST00000370132.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100212216.hg.1 |
NM_001918.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100212216.hg.1 |
TC0100015061.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300012031.hg.1 |
ENST00000496068.1 |
Ensembl |