Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02043128.hg.1 |
TC02002677.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02043128.hg.1 |
ENST00000315506.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02043128.hg.1 |
ENST00000315549.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02043128.hg.1 |
ENST00000359962.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02043128.hg.1 |
ENST00000374605.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02043128.hg.1 |
ENST00000396886.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02043128.hg.1 |
ENST00000409143.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02043128.hg.1 |
ENST00000409474.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02043128.hg.1 |
ENST00000428900.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02043128.hg.1 |
ENST00000447335.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02043128.hg.1 |
NM_033066.2 |
Refseq |