Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR07021118.hg.1 |
TC07001351.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07021118.hg.1 |
ENST00000258770.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07021118.hg.1 |
ENST00000361278.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07021118.hg.1 |
ENST00000395655.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07021118.hg.1 |
ENST00000471142.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07021118.hg.1 |
NM_004749.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07021118.hg.1 |
NM_030900.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07021118.hg.1 |
NM_199122.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07021141.hg.1 |
TC07001351.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07021141.hg.1 |
ENST00000361278.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07021141.hg.1 |
ENST00000395655.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07021141.hg.1 |
NM_004749.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07021141.hg.1 |
NM_030900.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07021141.hg.1 |
NM_199122.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07021140.hg.1 |
TC07001351.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07021140.hg.1 |
ENST00000258770.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07021140.hg.1 |
ENST00000361278.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07021140.hg.1 |
ENST00000395655.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07021140.hg.1 |
NM_004749.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07021140.hg.1 |
NM_030900.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07021140.hg.1 |
NM_199122.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700178288.hg.1 |
ENST00000258770.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700178288.hg.1 |
ENST00000361278.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700178288.hg.1 |
ENST00000395655.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700178288.hg.1 |
ENST00000461363.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700178288.hg.1 |
ENST00000471142.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700178288.hg.1 |
ENST00000475893.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700178288.hg.1 |
ENST00000478532.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700178288.hg.1 |
ENST00000482285.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700178288.hg.1 |
ENST00000495078.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700178288.hg.1 |
NM_004749.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700178288.hg.1 |
NM_030900.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700178288.hg.1 |
NM_199122.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700178288.hg.1 |
ENST00000258770.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700178288.hg.1 |
ENST00000361278.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700178288.hg.1 |
ENST00000395655.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700178288.hg.1 |
ENST00000461363.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700178288.hg.1 |
ENST00000471142.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700178288.hg.1 |
ENST00000475893.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700178288.hg.1 |
ENST00000478532.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700178288.hg.1 |
ENST00000482285.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700178288.hg.1 |
ENST00000495078.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700178288.hg.1 |
NM_004749.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700178288.hg.1 |
NM_030900.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700178288.hg.1 |
NM_199122.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700178288.hg.1 |
TC0700010947.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700178300.hg.1 |
ENST00000395655.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700178300.hg.1 |
NM_001261834.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700178300.hg.1 |
NM_004749.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700178300.hg.1 |
NM_030900.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700178300.hg.1 |
NM_199122.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700178300.hg.1 |
ENST00000395655.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700178300.hg.1 |
NM_001261834.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700178300.hg.1 |
NM_004749.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700178300.hg.1 |
NM_030900.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700178300.hg.1 |
NM_199122.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700178300.hg.1 |
TC0700010947.hg.1 |
Clariom D TC |