Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204218.1 | NP_001191147.1 | 17 - 18 | 3346 | 84 | |
XM_011538398.2 | XP_011536700.1 | 18 - 19 | 3217 | 84 | ||
XM_017019345.1 | XP_016874834.1 | 17 - 18 | 3353 | 84 | ||
XM_017019346.1 | XP_016874835.1 | 17 - 18 | 3167 | 84 | ||
GenBank mRNA | AJ004918.1 | - | 2 - 3 | 133 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Homo sapiens Probe spans exons |