Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007948.hg.1 |
ENST00000265462.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007948.hg.1 |
ENST00000265462.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007948.hg.1 |
ENST00000265462.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007948.hg.1 |
ENST00000265462.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007948.hg.1 |
ENST00000265462.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007948.hg.1 |
ENST00000265462.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007948.hg.1 |
ENST00000265462.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007948.hg.1 |
ENST00000265462.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007948.hg.1 |
ENST00000265462.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007948.hg.1 |
ENST00000265462.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007948.hg.1 |
ENST00000347941.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007948.hg.1 |
ENST00000347941.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007948.hg.1 |
ENST00000347941.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007948.hg.1 |
ENST00000347941.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007948.hg.1 |
ENST00000347941.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007948.hg.1 |
ENST00000347941.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007948.hg.1 |
ENST00000352435.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007948.hg.1 |
ENST00000352435.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007948.hg.1 |
ENST00000352435.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007948.hg.1 |
ENST00000352435.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007948.hg.1 |
ENST00000352435.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007948.hg.1 |
ENST00000352435.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007948.hg.1 |
ENST00000352435.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007948.hg.1 |
NM_012094.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007948.hg.1 |
NM_012094.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007948.hg.1 |
NM_012094.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007948.hg.1 |
NM_012094.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007948.hg.1 |
NM_012094.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007948.hg.1 |
NM_012094.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007948.hg.1 |
NM_012094.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007948.hg.1 |
NM_012094.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007948.hg.1 |
NM_012094.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007948.hg.1 |
NM_012094.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007948.hg.1 |
NM_181651.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007948.hg.1 |
NM_181651.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007948.hg.1 |
NM_181651.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007948.hg.1 |
NM_181651.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007948.hg.1 |
NM_181651.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007948.hg.1 |
NM_181651.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007948.hg.1 |
NM_181651.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007948.hg.1 |
NM_181651.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007948.hg.1 |
NM_181651.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007948.hg.1 |
NM_181652.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007948.hg.1 |
NM_181652.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007948.hg.1 |
NM_181652.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007948.hg.1 |
NM_181652.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007948.hg.1 |
NM_181652.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007948.hg.1 |
NM_181652.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007948.hg.1 |
NM_181652.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007948.hg.1 |
NM_181652.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11007342.hg.1 |
ENST00000265462.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11007342.hg.1 |
ENST00000347941.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11007342.hg.1 |
ENST00000352435.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11007342.hg.1 |
NM_012094.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11007342.hg.1 |
NM_181651.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11007342.hg.1 |
NM_181652.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11007342.hg.1 |
TC11000597.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11007343.hg.1 |
ENST00000265462.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11007343.hg.1 |
ENST00000347941.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11007343.hg.1 |
ENST00000352435.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11007343.hg.1 |
NM_012094.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11007343.hg.1 |
NM_181651.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11007343.hg.1 |
NM_181652.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11007343.hg.1 |
TC11000597.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1100007948.hg.1 |
ENST00000265462.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007948.hg.1 |
ENST00000265462.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007948.hg.1 |
ENST00000265462.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007948.hg.1 |
ENST00000265462.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007948.hg.1 |
ENST00000265462.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007948.hg.1 |
ENST00000265462.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007948.hg.1 |
ENST00000265462.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007948.hg.1 |
ENST00000265462.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007948.hg.1 |
ENST00000265462.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007948.hg.1 |
ENST00000265462.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007948.hg.1 |
ENST00000347941.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007948.hg.1 |
ENST00000347941.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007948.hg.1 |
ENST00000347941.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007948.hg.1 |
ENST00000347941.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007948.hg.1 |
ENST00000347941.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007948.hg.1 |
ENST00000347941.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007948.hg.1 |
ENST00000352435.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007948.hg.1 |
ENST00000352435.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007948.hg.1 |
ENST00000352435.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007948.hg.1 |
ENST00000352435.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007948.hg.1 |
ENST00000352435.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007948.hg.1 |
ENST00000352435.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007948.hg.1 |
ENST00000352435.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007948.hg.1 |
NM_012094.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007948.hg.1 |
NM_012094.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007948.hg.1 |
NM_012094.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007948.hg.1 |
NM_012094.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007948.hg.1 |
NM_012094.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007948.hg.1 |
NM_012094.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007948.hg.1 |
NM_012094.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007948.hg.1 |
NM_012094.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007948.hg.1 |
NM_012094.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007948.hg.1 |
NM_012094.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007948.hg.1 |
NM_181651.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007948.hg.1 |
NM_181651.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007948.hg.1 |
NM_181651.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007948.hg.1 |
NM_181651.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007948.hg.1 |
NM_181651.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007948.hg.1 |
NM_181651.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007948.hg.1 |
NM_181651.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007948.hg.1 |
NM_181651.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007948.hg.1 |
NM_181651.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007948.hg.1 |
NM_181652.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007948.hg.1 |
NM_181652.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007948.hg.1 |
NM_181652.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007948.hg.1 |
NM_181652.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007948.hg.1 |
NM_181652.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007948.hg.1 |
NM_181652.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007948.hg.1 |
NM_181652.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007948.hg.1 |
NM_181652.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100157058.hg.1 |
ENST00000265462.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100157058.hg.1 |
ENST00000347941.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100157058.hg.1 |
ENST00000352435.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100157058.hg.1 |
NM_012094.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100157058.hg.1 |
NM_181651.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100157058.hg.1 |
NM_181652.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100157058.hg.1 |
ENST00000265462.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100157058.hg.1 |
ENST00000347941.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100157058.hg.1 |
ENST00000352435.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100157058.hg.1 |
NM_012094.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100157058.hg.1 |
NM_181651.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100157058.hg.1 |
NM_181652.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100157058.hg.1 |
TC1100007948.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100157057.hg.1 |
ENST00000265462.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100157057.hg.1 |
ENST00000347941.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100157057.hg.1 |
ENST00000352435.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100157057.hg.1 |
NM_012094.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100157057.hg.1 |
NM_181651.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100157057.hg.1 |
NM_181652.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100157057.hg.1 |
ENST00000265462.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100157057.hg.1 |
ENST00000347941.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100157057.hg.1 |
ENST00000352435.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100157057.hg.1 |
NM_012094.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100157057.hg.1 |
NM_181651.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100157057.hg.1 |
NM_181652.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100157057.hg.1 |
TC1100007948.hg.1 |
Clariom D TC |