Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11024976.hg.1 |
ENST00000310128.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11024976.hg.1 |
ENST00000525550.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11024976.hg.1 |
NM_005472.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11024976.hg.1 |
TC11002082.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11024980.hg.1 |
ENST00000310128.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11024980.hg.1 |
NM_005472.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11024980.hg.1 |
TC11002082.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11024975.hg.1 |
ENST00000310128.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11024975.hg.1 |
ENST00000525550.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11024975.hg.1 |
NM_005472.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11024975.hg.1 |
TC11002082.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011614.hg.1 |
ENST00000310128.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011614.hg.1 |
ENST00000310128.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011614.hg.1 |
ENST00000310128.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011614.hg.1 |
ENST00000310128.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011614.hg.1 |
ENST00000310128.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011614.hg.1 |
ENST00000310128.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011614.hg.1 |
ENST00000310128.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011614.hg.1 |
ENST00000310128.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011614.hg.1 |
ENST00000310128.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011614.hg.1 |
ENST00000310128.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011614.hg.1 |
ENST00000310128.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011614.hg.1 |
ENST00000310128.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011614.hg.1 |
ENST00000525550.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011614.hg.1 |
ENST00000525550.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011614.hg.1 |
ENST00000525550.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011614.hg.1 |
ENST00000525550.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011614.hg.1 |
ENST00000525550.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011614.hg.1 |
ENST00000525550.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011614.hg.1 |
ENST00000525550.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011614.hg.1 |
ENST00000525550.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011614.hg.1 |
ENST00000526855.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011614.hg.1 |
ENST00000526855.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011614.hg.1 |
ENST00000526855.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011614.hg.1 |
ENST00000526855.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011614.hg.1 |
ENST00000526855.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011614.hg.1 |
ENST00000526855.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011614.hg.1 |
ENST00000526855.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011614.hg.1 |
ENST00000529425.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011614.hg.1 |
ENST00000529425.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011614.hg.1 |
ENST00000529425.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011614.hg.1 |
ENST00000529425.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011614.hg.1 |
ENST00000529425.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011614.hg.1 |
ENST00000529425.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011614.hg.1 |
ENST00000529425.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011614.hg.1 |
ENST00000531854.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011614.hg.1 |
ENST00000531854.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011614.hg.1 |
ENST00000531854.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011614.hg.1 |
ENST00000531854.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011614.hg.1 |
ENST00000531854.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011614.hg.1 |
ENST00000531854.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011614.hg.1 |
ENST00000531854.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011614.hg.1 |
ENST00000532569.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011614.hg.1 |
ENST00000532569.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011614.hg.1 |
ENST00000532569.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011614.hg.1 |
ENST00000532569.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011614.hg.1 |
ENST00000532569.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011614.hg.1 |
ENST00000532569.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011614.hg.1 |
ENST00000532569.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011614.hg.1 |
NM_005472.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011614.hg.1 |
NM_005472.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011614.hg.1 |
NM_005472.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011614.hg.1 |
NM_005472.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011614.hg.1 |
NM_005472.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011614.hg.1 |
NM_005472.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011614.hg.1 |
NM_005472.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011614.hg.1 |
NM_005472.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011614.hg.1 |
NM_005472.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011614.hg.1 |
NM_005472.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011614.hg.1 |
NM_005472.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100189692.hg.1 |
ENST00000310128.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100189692.hg.1 |
ENST00000525550.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100189692.hg.1 |
ENST00000526855.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100189692.hg.1 |
ENST00000529425.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100189692.hg.1 |
ENST00000531854.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100189692.hg.1 |
ENST00000532569.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100189692.hg.1 |
NM_005472.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100189692.hg.1 |
ENST00000310128.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100189692.hg.1 |
ENST00000525550.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100189692.hg.1 |
ENST00000526855.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100189692.hg.1 |
ENST00000529425.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100189692.hg.1 |
ENST00000531854.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100189692.hg.1 |
ENST00000532569.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100189692.hg.1 |
NM_005472.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100189692.hg.1 |
TC1100011614.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100189693.hg.1 |
ENST00000310128.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100189693.hg.1 |
ENST00000525550.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100189693.hg.1 |
ENST00000526855.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100189693.hg.1 |
ENST00000529425.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100189693.hg.1 |
ENST00000531854.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100189693.hg.1 |
ENST00000532569.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100189693.hg.1 |
NM_005472.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100189693.hg.1 |
ENST00000310128.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100189693.hg.1 |
ENST00000525550.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100189693.hg.1 |
ENST00000526855.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100189693.hg.1 |
ENST00000529425.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100189693.hg.1 |
ENST00000531854.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100189693.hg.1 |
ENST00000532569.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100189693.hg.1 |
NM_005472.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100189693.hg.1 |
TC1100011614.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100189698.hg.1 |
ENST00000310128.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100189698.hg.1 |
ENST00000526855.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100189698.hg.1 |
ENST00000529425.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100189698.hg.1 |
ENST00000531854.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100189698.hg.1 |
NM_005472.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100189698.hg.1 |
ENST00000310128.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100189698.hg.1 |
ENST00000526855.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100189698.hg.1 |
ENST00000529425.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100189698.hg.1 |
ENST00000531854.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100189698.hg.1 |
NM_005472.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100189698.hg.1 |
TC1100011614.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100011614.hg.1 |
ENST00000310128.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011614.hg.1 |
ENST00000310128.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011614.hg.1 |
ENST00000310128.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011614.hg.1 |
ENST00000310128.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011614.hg.1 |
ENST00000310128.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011614.hg.1 |
ENST00000310128.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011614.hg.1 |
ENST00000310128.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011614.hg.1 |
ENST00000310128.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011614.hg.1 |
ENST00000310128.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011614.hg.1 |
ENST00000310128.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011614.hg.1 |
ENST00000310128.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011614.hg.1 |
ENST00000310128.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011614.hg.1 |
ENST00000525550.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011614.hg.1 |
ENST00000525550.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011614.hg.1 |
ENST00000525550.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011614.hg.1 |
ENST00000525550.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011614.hg.1 |
ENST00000525550.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011614.hg.1 |
ENST00000525550.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011614.hg.1 |
ENST00000525550.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011614.hg.1 |
ENST00000525550.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011614.hg.1 |
ENST00000526855.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011614.hg.1 |
ENST00000526855.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011614.hg.1 |
ENST00000526855.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011614.hg.1 |
ENST00000526855.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011614.hg.1 |
ENST00000526855.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011614.hg.1 |
ENST00000526855.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011614.hg.1 |
ENST00000526855.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011614.hg.1 |
ENST00000529425.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011614.hg.1 |
ENST00000529425.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011614.hg.1 |
ENST00000529425.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011614.hg.1 |
ENST00000529425.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011614.hg.1 |
ENST00000529425.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011614.hg.1 |
ENST00000529425.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011614.hg.1 |
ENST00000529425.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011614.hg.1 |
ENST00000531854.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011614.hg.1 |
ENST00000531854.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011614.hg.1 |
ENST00000531854.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011614.hg.1 |
ENST00000531854.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011614.hg.1 |
ENST00000531854.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011614.hg.1 |
ENST00000531854.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011614.hg.1 |
ENST00000531854.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011614.hg.1 |
ENST00000532569.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011614.hg.1 |
ENST00000532569.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011614.hg.1 |
ENST00000532569.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011614.hg.1 |
ENST00000532569.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011614.hg.1 |
ENST00000532569.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011614.hg.1 |
ENST00000532569.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011614.hg.1 |
ENST00000532569.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011614.hg.1 |
NM_005472.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011614.hg.1 |
NM_005472.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011614.hg.1 |
NM_005472.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011614.hg.1 |
NM_005472.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011614.hg.1 |
NM_005472.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011614.hg.1 |
NM_005472.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011614.hg.1 |
NM_005472.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011614.hg.1 |
NM_005472.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011614.hg.1 |
NM_005472.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011614.hg.1 |
NM_005472.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011614.hg.1 |
NM_005472.4 |
Refseq |