Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR20010571.hg.1 |
ENST00000252015.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20010571.hg.1 |
ENST00000432634.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20010571.hg.1 |
ENST00000451813.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20010571.hg.1 |
ENST00000541994.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20010571.hg.1 |
NM_015638.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20010571.hg.1 |
NM_199368.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20010571.hg.1 |
TC20000791.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2000162359.hg.1 |
ENST00000252015.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000162359.hg.1 |
ENST00000451813.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000162359.hg.1 |
NM_015638.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000162359.hg.1 |
NM_199368.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000162359.hg.1 |
TC2000008955.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000162359.hg.1 |
ENST00000252015.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000162359.hg.1 |
ENST00000451813.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000162359.hg.1 |
NM_015638.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000162359.hg.1 |
NM_199368.1 |
Refseq |