Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008946.hg.1 |
ENST00000336431.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008946.hg.1 |
ENST00000336431.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008946.hg.1 |
ENST00000336431.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008946.hg.1 |
ENST00000336431.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008946.hg.1 |
ENST00000336431.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008946.hg.1 |
ENST00000336431.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008946.hg.1 |
ENST00000336431.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008946.hg.1 |
ENST00000336431.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008946.hg.1 |
ENST00000336431.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008946.hg.1 |
ENST00000336431.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008946.hg.1 |
ENST00000336431.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008946.hg.1 |
ENST00000336431.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008946.hg.1 |
ENST00000336431.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008946.hg.1 |
ENST00000336431.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008946.hg.1 |
ENST00000336431.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008946.hg.1 |
ENST00000336431.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008946.hg.1 |
ENST00000336431.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008946.hg.1 |
ENST00000336431.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008946.hg.1 |
ENST00000336431.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008946.hg.1 |
ENST00000336431.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008946.hg.1 |
ENST00000427420.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008946.hg.1 |
ENST00000427420.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008946.hg.1 |
ENST00000427420.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008946.hg.1 |
ENST00000427420.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008946.hg.1 |
ENST00000469018.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008946.hg.1 |
ENST00000469018.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008946.hg.1 |
ENST00000469018.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008946.hg.1 |
ENST00000469018.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008946.hg.1 |
ENST00000469018.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008946.hg.1 |
ENST00000469018.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008946.hg.1 |
ENST00000469018.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008946.hg.1 |
ENST00000469018.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008946.hg.1 |
ENST00000470952.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008946.hg.1 |
ENST00000470952.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008946.hg.1 |
ENST00000470952.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008946.hg.1 |
ENST00000470952.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008946.hg.1 |
NM_178026.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008946.hg.1 |
NM_178026.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008946.hg.1 |
NM_178026.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008946.hg.1 |
NM_178026.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008946.hg.1 |
NM_178026.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008946.hg.1 |
NM_178026.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008946.hg.1 |
NM_178026.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008946.hg.1 |
NM_178026.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008946.hg.1 |
NM_178026.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008946.hg.1 |
NM_178026.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008946.hg.1 |
NM_178026.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008946.hg.1 |
NM_178026.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008946.hg.1 |
NM_178026.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008946.hg.1 |
NM_178026.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008946.hg.1 |
NM_178026.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008946.hg.1 |
NM_178026.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008946.hg.1 |
NM_178026.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008946.hg.1 |
NM_178026.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008946.hg.1 |
NM_178026.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008946.hg.1 |
NM_178026.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20010516.hg.1 |
ENST00000336431.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20010516.hg.1 |
NM_178026.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20010516.hg.1 |
TC20000788.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2000162298.hg.1 |
ENST00000336431.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000162298.hg.1 |
NM_178026.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000162298.hg.1 |
TC2000008946.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000162298.hg.1 |
ENST00000336431.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000162298.hg.1 |
NM_178026.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008946.hg.1 |
ENST00000336431.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008946.hg.1 |
ENST00000336431.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008946.hg.1 |
ENST00000336431.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008946.hg.1 |
ENST00000336431.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008946.hg.1 |
ENST00000336431.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008946.hg.1 |
ENST00000336431.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008946.hg.1 |
ENST00000336431.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008946.hg.1 |
ENST00000336431.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008946.hg.1 |
ENST00000336431.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008946.hg.1 |
ENST00000336431.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008946.hg.1 |
ENST00000336431.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008946.hg.1 |
ENST00000336431.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008946.hg.1 |
ENST00000336431.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008946.hg.1 |
ENST00000336431.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008946.hg.1 |
ENST00000336431.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008946.hg.1 |
ENST00000336431.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008946.hg.1 |
ENST00000336431.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008946.hg.1 |
ENST00000336431.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008946.hg.1 |
ENST00000336431.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008946.hg.1 |
ENST00000336431.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008946.hg.1 |
ENST00000427420.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008946.hg.1 |
ENST00000427420.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008946.hg.1 |
ENST00000427420.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008946.hg.1 |
ENST00000427420.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008946.hg.1 |
ENST00000469018.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008946.hg.1 |
ENST00000469018.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008946.hg.1 |
ENST00000469018.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008946.hg.1 |
ENST00000469018.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008946.hg.1 |
ENST00000469018.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008946.hg.1 |
ENST00000469018.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008946.hg.1 |
ENST00000469018.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008946.hg.1 |
ENST00000469018.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008946.hg.1 |
ENST00000470952.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008946.hg.1 |
ENST00000470952.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008946.hg.1 |
ENST00000470952.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008946.hg.1 |
ENST00000470952.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008946.hg.1 |
NM_178026.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008946.hg.1 |
NM_178026.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008946.hg.1 |
NM_178026.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008946.hg.1 |
NM_178026.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008946.hg.1 |
NM_178026.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008946.hg.1 |
NM_178026.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008946.hg.1 |
NM_178026.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008946.hg.1 |
NM_178026.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008946.hg.1 |
NM_178026.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008946.hg.1 |
NM_178026.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008946.hg.1 |
NM_178026.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008946.hg.1 |
NM_178026.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008946.hg.1 |
NM_178026.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008946.hg.1 |
NM_178026.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008946.hg.1 |
NM_178026.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008946.hg.1 |
NM_178026.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008946.hg.1 |
NM_178026.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008946.hg.1 |
NM_178026.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008946.hg.1 |
NM_178026.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008946.hg.1 |
NM_178026.2 |
Refseq |