Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Clariom D Assay |
PSR0900185206.hg.1 |
ENST00000371757.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900185206.hg.1 |
ENST00000460750.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900185206.hg.1 |
ENST00000475008.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900185206.hg.1 |
ENST00000486577.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900185206.hg.1 |
ENST00000487664.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900185206.hg.1 |
ENST00000488444.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900185206.hg.1 |
ENST00000490355.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900185206.hg.1 |
ENST00000491806.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900185206.hg.1 |
ENST00000630792.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900185206.hg.1 |
ENST00000631073.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900185206.hg.1 |
NM_001272003.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900185206.hg.1 |
NM_020822.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900185206.hg.1 |
ENST00000371757.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900185206.hg.1 |
ENST00000460750.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900185206.hg.1 |
ENST00000475008.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900185206.hg.1 |
ENST00000486577.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900185206.hg.1 |
ENST00000487664.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900185206.hg.1 |
ENST00000488444.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900185206.hg.1 |
ENST00000490355.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900185206.hg.1 |
ENST00000491806.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900185206.hg.1 |
ENST00000630792.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900185206.hg.1 |
ENST00000631073.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900185206.hg.1 |
NM_001272003.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900185206.hg.1 |
NM_020822.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900185206.hg.1 |
TC0900012187.hg.1 |
Clariom D TC |